Skip to Content
Merck
All Photos(1)

Key Documents

106577

Sigma-Aldrich

2-Iodohippuric acid

98%

Synonym(s):

N-(2-Iodobenzoyl)glycine

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$130.00

$130.00


Estimated to ship onApril 10, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$130.00

About This Item

Linear Formula:
IC6H4CONHCH2CO2H
CAS Number:
Molecular Weight:
305.07
Beilstein:
1877943
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

$130.00


Estimated to ship onApril 10, 2025


Request a Bulk Order

Quality Level

Assay

98%

reaction suitability

reaction type: solution phase peptide synthesis

mp

170-172 °C (lit.)

SMILES string

OC(=O)CNC(=O)c1ccccc1I

InChI

1S/C9H8INO3/c10-7-4-2-1-3-6(7)9(14)11-5-8(12)13/h1-4H,5H2,(H,11,14)(H,12,13)

InChI key

CORFWQGVBFFZHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W M Michels et al.
Clinical nephrology, 72(5), 337-343 (2009-10-30)
Measuring GFR using exogenous markers without a bladder catheter, errors can be easily made due to incomplete urine collection. The aim was to quantify agreement for 125I-iothalamate GFR measurements with and without a correction for inaccurate urine collection using 131I-hippuran.
Andrew T Taylor et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(4), 578-584 (2013-02-21)
The pharmacokinetics of the tricarbonyl core radiopharmaceutical (99m)Tc(CO)3-nitrilotriacetic acid ((99m)Tc(CO)3(NTA)) in rats and subjects with normal renal function are comparable to those of (131)I-o-iodohippuran ((131)I-OIH), the radiopharmaceutical gold standard for the measurement of effective renal plasma flow. Our objective was
Andrew T Taylor et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 45(5), 885-891 (2004-05-12)
99mTc-Mercaptoacetyltriglycine ((99m)Tc-MAG3) and (99m)Tc-L,L-ethylenedicysteine ((99m)Tc-LL-EC) are useful renal radiopharmaceuticals; however, both agents have renal clearances less than that of (131)I-orthoiodohippurate ((131)I-OIH), and (99m)Tc-LL-EC exists in dianionic and monoanionic forms at physiologic pH. In an effort to develop a superior (99m)Tc
Gopal Pathuri et al.
Nuclear medicine communications, 32(10), 908-912 (2011-08-31)
We recently identified p-[(18)F]fluorohippurate ([(18)F]PFH) as a potential positron emission tomography (PET) renal agent. The objective of this study was to compare renogram parameters of [(18)F]PFH with o-[(125)I]iodohippurate ([(125)I]OIH) as a surrogate for the renal imaging gold standard (131)I-OIH and
Raymond Miralbell et al.
International journal of radiation oncology, biology, physics, 58(3), 809-816 (2004-02-18)
Patients with malignant hematologic disorders undergoing bone marrow transplantation (BMT) may develop renal insufficiency. A study was undertaken to assess prospectively the subclinical renal function changes with radioisotopic methods in patients undergoing BMT for hematologic malignancies. We studied 71 patients

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service