Skip to Content
Merck
All Photos(1)

Key Documents

103314

Sigma-Aldrich

Chloro(4-chlorophenyl)phenylmethane

97%

Synonym(s):

4-Chlorobenzhydryl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH(C6H5)Cl
CAS Number:
Molecular Weight:
237.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.603 (lit.)

bp

159-160 °C/2 mmHg (lit.)

density

1.239 g/mL at 25 °C (lit.)

SMILES string

ClC(c1ccccc1)c2ccc(Cl)cc2

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Contact dermatitis due to p-chlorophenyl phenyl chloromethane (author's transl)].
A Rothe et al.
Dermatologische Monatschrift, 166(6), 389-393 (1980-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service