Skip to Content
Merck
All Photos(1)

Key Documents

103292

Sigma-Aldrich

6-Methoxyquinaldine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO
CAS Number:
Molecular Weight:
173.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

62-64 °C (lit.)

SMILES string

COc1ccc2nc(C)ccc2c1

InChI

1S/C11H11NO/c1-8-3-4-9-7-10(13-2)5-6-11(9)12-8/h3-7H,1-2H3

InChI key

NAGJQQFMJKMXJQ-UHFFFAOYSA-N

General description

6-Methoxyquinaldine is starting reagent for the synthesis of 2-(-dialkylaminoethyl)-1,2,3,4-tetrahydroquinolines and their derivatives[1].

Application

6-Methoxyquinaldine was used in the preparation and characterization of three RNA-specific fluorescent probes (E36, E144and F22) and their applications in live cell imaging[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quinidines substituted 1, 1, 2, 3-tetrahydroquinolines.
Shchukina MN, et al.
Pharmaceutical Chemistry Journal, 9(2), 94-97 (1975)
Qian Li et al.
Nature protocols, 1(6), 2922-2932 (2007-04-05)
This protocol outlines a methodology for the preparation and characterization of three RNA-specific fluorescent probes (E36, E144 and F22) and their use in live cell imaging. It describes a detailed procedure for their chemical synthesis and purification; serial product characterization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service