Skip to Content
Merck
All Photos(1)

Documents

100781

Sigma-Aldrich

Benzyl methyl sulfide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H10S
CAS Number:
Molecular Weight:
138.23
EC Number:
UNSPSC Code:
12352100

Assay

98%

refractive index

n20/D 1.562 (lit.)

bp

195-198 °C (lit.)

density

1.015 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(cc1)C(=O)c2ccccc2

InChI

1S/C14H12O/c1-11-7-9-13(10-8-11)14(15)12-5-3-2-4-6-12/h2-10H,1H3

InChI key

WXPWZZHELZEVPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service