Skip to Content
Merck
All Photos(1)

Key Documents

100307

Sigma-Aldrich

2,6-Dichlorobenzyl bromide

97%

Synonym(s):

α-Bromo-2,6-dichlorotoluene, 1-Bromomethyl-2,6-dichlorobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$40.40

$40.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$40.40

About This Item

Linear Formula:
Cl2C6H3CH2Br
CAS Number:
Molecular Weight:
239.92
Beilstein:
637351
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$40.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

54-56 °C (lit.)

functional group

bromo
chloro

SMILES string

Clc1cccc(Cl)c1CBr

InChI

1S/C7H5BrCl2/c8-4-5-6(9)2-1-3-7(5)10/h1-3H,4H2

InChI key

PDFGFQUSSYSWNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service