Skip to Content
Merck
All Photos(3)

Key Documents

100242

Sigma-Aldrich

3-Aminophenol

98%

Synonym(s):

m-Aminophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4OH
CAS Number:
Molecular Weight:
109.13
Beilstein:
636059
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

solid

bp

164 °C/11 mmHg (lit.)

mp

120-124 °C (lit.)

SMILES string

Nc1cccc(O)c1

InChI

1S/C6H7NO/c7-5-2-1-3-6(8)4-5/h1-4,8H,7H2

InChI key

CWLKGDAVCFYWJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Aminophenol has been used in the synthesis of disulfonated bis[4-(3-aminophenoxy)phenyl]sulfone (S-BAPS).[1]
It can be used to synthesize:
  • Methyl 2-oxo-7-[(triphenylphosphoranylidene)amino]-2H-chromene-4-carboxylate by reacting with dimethyl acetylenedicarboxylate (DMAD) in the presence of triphenylphosphine.[2]
  • 3-Amino-2-cyclohexen-1-one via palladium-catalyzed hydrogenation.[3]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sijing Chen et al.
Polymers, 11(6) (2019-06-20)
Benzoxazine containing fluorinated aromatic ether nitrile linkage (FAEN-Bz) had been synthesized from 2,6-dichlorobenzonitrile, 4,4'-(hexafluoroisopropylidene)diphenol (bisphenol AF), 3-Aminophenol, formaldehyde, phenol by condensation polymerization and Mannich ring-forming reaction. Structures of the monomer were verified by Proton NMR spectrum (1H-NMR) and Fourier transform
A Practical and One-Pot Procedure for the Synthesis of 3-Amino-2-cyclohexen-1-one from 3-Aminophenol.
Sajiki H
Organic Process Research & Development, 9(2), 219-220 (2005)
Vinyltriphenylphosphonium salt mediated synthesis of 1, 4-benzoxazine and coumarin derivatives.
Yavari I, et al.
Tetrahedron, 58(34), 6895-6899 (2002)
Ping-Chih Hsu et al.
Oncology reports, 42(2), 697-707 (2019-06-25)
Human non‑small cell lung cancer (NSCLC) is associated with an extremely poor prognosis especially for the 40% of patients who develop brain metastasis, and few treatment strategies exist. Cucurbitacin E (CuE), an oxygenated tetracyclic triterpenoid isolated from plants particularly of
Simultaneous spectrophotometric determination of PAS and its degradation product m-aminophenol.
C Vetuschi et al.
Pharmaceutica acta Helvetiae, 63(11), 290-293 (1988-01-01)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service