Skip to Content
Merck
All Photos(1)

Key Documents

05656

Sigma-Aldrich

N-[4-(3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl)benzyloxycarbonyloxy] succinimide

≥97%

Synonym(s):

N-[4-(1H,1H,2H,2H-Perfluorooctyl)benzyloxycarbonyloxy]succinimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14F13NO5
CAS Number:
Molecular Weight:
595.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97%

mp

113-118 °C

storage temp.

−20°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCc1ccc(COC(=O)ON2C(=O)CCC2=O)cc1

InChI

1S/C20H14F13NO5/c21-15(22,16(23,24)17(25,26)18(27,28)19(29,30)20(31,32)33)8-7-10-1-3-11(4-2-10)9-38-14(37)39-34-12(35)5-6-13(34)36/h1-4H,5-9H2

InChI key

XLFQCGUSTBBBAK-UHFFFAOYSA-N

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service