Skip to Content
Merck
All Photos(1)

Key Documents

T3566

Sigma-Aldrich

Clear-view Snap-Cap microtubes

size 1.5 mL, natural

Synonym(s):

low retention microtube, snap-cap vial

Sign Into View Organizational & Contract Pricing

Select a Size

500 EA
$32.80

$32.80


Estimated to ship onApril 09, 2025



Select a Size

Change View
500 EA
$32.80

About This Item

UNSPSC Code:
41121702
NACRES:
NB.11

$32.80


Estimated to ship onApril 09, 2025


material

cap (Angled cap lid)
polypropylene

feature

RNase and DNase free

packaging

pack of 500 ea

manufacturer/tradename

Excel Scientific MCT-1.5N

parameter

-20,000 × g max. RCF

size

1.5 mL

color

natural

Looking for similar products? Visit Product Comparison Guide

General description

  • Superior clarity for sample viewing
  • A consistent, ring-lock seal prevents inadvertent opening
  • Angled cap lid makes opening easy without thumb fatigue
  • Large flat tops for ease of identification and a pierceable cap
  • Large, crisp reference lines and graduation numbers
  • Certified RNase/DNase free

Legal Information

Clear-view is a trademark of Excel Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service