Skip to Content
Merck
All Photos(5)

Key Documents

G5882

Sigma-Aldrich

Glutaraldehyde solution

Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative

Synonym(s):

Glutaric dialdehyde solution, Pentane-1,5-dial

Sign Into View Organizational & Contract Pricing

Select a Size

10 X 1 ML
$222.00
50 ML
$250.00
80 ML
$301.00
10 X 10 ML
$323.00
100 ML
$338.00
1 L
$2,450.00

$222.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
10 X 1 ML
$222.00
50 ML
$250.00
80 ML
$301.00
10 X 10 ML
$323.00
100 ML
$338.00
1 L
$2,450.00

About This Item

Linear Formula:
OHC(CH2)3CHO
CAS Number:
Molecular Weight:
100.12
Beilstein:
605390
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.21

$222.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Quality Level

type

Grade I

form

liquid

concentration

25% in H2O

technique(s)

electron microscopy: suitable (fixative)
electron microscopy: suitable

color

colorless

useful pH range

2.9

mp

-10 °C ((14 °F ))

solubility

water: soluble at 20 °C

application(s)

cell analysis

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(CCCC([H])=O)=O

InChI

1S/C5H8O2/c6-4-2-1-3-5-7/h4-5H,1-3H2

InChI key

SXRSQZLOMIGNAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glutaraldehyde (Glutaric dialdehyde, Pentane-1,5-dial) is an aliphatic dialdehyde widely utilized as a versatile biochemical reagent. Glutaraldehyde solutions contain both monomers and polymers with the difference in their fixation properties. Its applications include serving as a protein crosslinker, facilitating enzyme immobilization in microscopy, histochemistry, and cytochemistry. This compound acts as an amine-reactive homobifunctional crosslinker and a cell fixative before SDS-PAGE, staining, or electron microscopy, making it suitable for diverse morphological studies.
Glutaraldehyde efficiently crosslinks amine and hydrazine derivatives to proteins and other amine-containing polymers. Notably, it has been used in certain reactions to directly couple biotin hydrazides to nucleic acids, suggesting potential applications in conjugating fluorescent hydrazides and hydroxylamines to DNA. In histology studies, Glutaraldehyde is employed to preserve tissue sections, preparing them for closer examination.

Application

Glutaraldehyde solution has been used:
  • to fix the blood samples for scanning electron microscopy (SEM)
  • to fix scaffold pieces for SEM analysis
  • in drug treatments to treat K562 (leukemia) cells to permanently cross-link cell components

Biochem/physiol Actions

Glutaraldehyde can be used in biomedical research to fix cells. It can also be used as a tool to investigate healthy and pathologic red blood cells. Medical equipment like spirometry tubing, endoscopes, dialyzers, hemodialysis proportioning and dialysate delivery systems, endocavitary probes, transducers, anesthesia and respiratory therapy equipment, and reuse of laparoscopic disposable plastic trocars can be disinfected with glutaraldehyde.

Features and Benefits

  • Versatile and adaptable for wide variety of research applications
  • Ready-made solution reduces the need for preparation time

Other Notes

For additional information on our range of Biochemicals, please complete this form.
Recommended for techniques demanding high purity.
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Malwina Sosnowska et al.
International journal of nanomedicine, 14, 6197-6215 (2019-09-10)
Degradation of the extracellular matrix (ECM) changes the physicochemical properties and dysregulates ECM-cell interactions, leading to several pathological conditions, such as invasive cancer. Carbon nanofilm, as a biocompatible and easy to functionalize material, could be used to mimic ECM structures
Jules D P Valentin et al.
Journal of colloid and interface science, 552, 247-257 (2019-05-28)
Many materials used in the medical settings such as catheters and contact lenses as well as most biological tissues are not purely elastic, but rather viscoelastic. While substrate elasticity has been investigated for its influence on bacterial adhesion, the impact
Mariafrancesca Cascione et al.
International journal of molecular sciences, 20(14) (2019-07-26)
The side effects induced by nanoparticle exposure at a cellular level are one of the priority research topics due to the steady increase in the use of nanoparticles (NPs). Recently, the focus on cellular morphology and mechanical behavior is gaining
Irene Costantini et al.
Scientific reports, 5, 9808-9808 (2015-05-08)
Extensive mapping of neuronal connections in the central nervous system requires high-throughput µm-scale imaging of large volumes. In recent years, different approaches have been developed to overcome the limitations due to tissue light scattering. These methods are generally developed to
Stéphane Jauréguiberry et al.
Blood, 124(2), 167-175 (2014-05-27)
Patients with severe malaria treated with artesunate sometimes experience a delayed hemolytic episode. Artesunate (AS) induces pitting, a splenic process whereby dead parasites are expelled from their host erythrocytes. These once-infected erythrocytes then return to the circulation. We analyzed hematologic

Related Content

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Is this concentration of 25% expressed in terms of weight by weight (w/w), weight by volume (w/v), or volume by volume (v/v)?

    1 answer
    1. The concentration of the product is 25% w/w solution.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service