Skip to Content
Merck
All Photos(1)

Key Documents

11663925910

Roche

Mycoplasma PCR ELISA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

usage

sufficient for 96 reactions

manufacturer/tradename

Roche

shipped in

dry ice

storage temp.

−20°C

Specificity

The Mycoplasma PCR ELISA is known to detect all of the following mycoplasma, acholeplasma, and ureaplasma species: M. orale, M. arginini, M. fermentans, M. hyorhinis, M. salivarium, M. gallisepticum, M. hominis, M. bovis, M. californicum, M. bovoculi, M. Pg50 bovine group, M. bovigenitalium, M. hyopneumoniae, A. laidlawii, U. urealyticum. DNA from other bacteria (except for Clostridium sp.), yeast, and eukaryotic cells are not detected.

Application

For research use only. Not for use in diagnostic procedures.
Mycoplasma PCR ELISA kit has been used for the detection of contamination by mycoplasma/acholeplasma/ureaplasma in cell cultures or other sample materials.
Mycoplasma PCR ELISA has been used to test mycoplasma contamination in cell lines.

Features and Benefits

Assay time: approximately 6 hours
Sample material: Cell culture supernatants, cell suspensions, or other samples
Sensitivity: At least 1 x 103 colony-forming units (cfu) of mycoplasma per milliliter of culture medium
  • High sensitivity: Detects 1 - 10fg of DNA, corresponding to approximately 1 - 20 gene copies per reaction (at least 103cfu/ml)
  • Broad detection range: All mycoplasma, acholeplasma, and ureaplasma species tested thus far were detected
  • Simplicity: Detects all mycoplasma using the same protocol, as opposed to the culture method that requires different culture conditions for different mycoplasma species
  • Fast results: The entire procedure can be performed in one day
  • Easy handling of large numbers of samples: The ready-to-use mix and ELISA detection procedure allow fast and semi-automatic sample processing

Packaging

1 kit containing 14 components.

Sequence

Chain Length Positive control: 520 bp.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Lysis Reagent

  • Neutralization Reagent

  • PCR Mix ready-to-use

  • Denaturation Reagent

  • Hybridization Buffer

  • Biotinylated Capture Probe

  • Washing Buffer 10x concentrated

  • Anti-Digoxigenin-POD antibody

  • Conjugate Dilution Buffer

  • TMB Substrate Solution

  • Stop Reagent

  • Positive Control DNA

  • Streptavidin-coated Microplate

  • Adhesive Plate Cover Foils

See All (14)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Novel Non-phosphorylated Serine 9/21 GSK3?/? Antibodies: Expanding the Tools for Studying GSK3 Regulation
Grabinski T, et al.
Frontiers in Molecular Neuroscience, 9, 123-123 (2016)
Kunal Karve et al.
Cell death & disease, 11(8), 704-704 (2020-08-28)
Alternative splicing contributes to diversification of gene function, yet consequences of splicing on functions of specific gene products is poorly understood. The major transcription factor TCF7L2 undergoes alternative splicing but the biological significance of TCF7L2 isoforms has remained largely to
Pengfei Wang et al.
Nature, 593(7857), 130-135 (2021-03-09)
The COVID-19 pandemic has had widespread effects across the globe, and its causative agent, SARS-CoV-2, continues to spread. Effective interventions need to be developed to end this pandemic. Single and combination therapies with monoclonal antibodies have received emergency use authorization1-3
Novel Non-phosphorylated Serine 9/21 GSK3?/a Antibodies: Expanding the Tools for Studying GSK3 Regulation
Tessa G, et al.
Frontiers in Molecular Neuroscience (2016)
Melanocytes Affect Nodal Expression and Signaling in Melanoma Cells: A Lesson from Pediatric Large Congenital Melanocytic Nevi
Margaryan N V, et al.
International Journal of Molecular Sciences, 17(3), 418-418 (2016)

Protocols

Mycoplasma contamination of cell cultures is a serious issue impacting cell model validity. PCR testing for mycoplasma is an inexpensive, sensitive, and specific method for detecting contamination.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service