Skip to Content
Merck
All Photos(1)

Key Documents

W387509

Sigma-Aldrich

Dimethyl sulfoxide

≥99%

Synonym(s):

DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
FEMA Number:
3875
Beilstein:
506008
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
Flavis number:
12.175

biological source

synthetic

grade

Halal

Agency

meets purity specifications of JECFA

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

573 °F

shelf life

Expiration date period - 5 years

expl. lim.

42 %, 63 °F

color

colorless

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)
189 °C

mp

16-19 °C (lit.)

solubility

water: miscible

density

1.10 g/mL (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

butter; sulfurous

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sulfoxide (DMSO) is a hygroscopic, high polarity amphiphilic solvent with an ability to dissolve many compounds. Cytotoxic study of DMSO on the pulp tissue repair-related activity of odontoblast-like cells showed it produced minor effects. Along with phosphorus pentaoxide it participates in the oxidation of carbohydrates to afford aldosuloses and aldosiduloses. DMSO participates in the oxidation of some partially-acetylated carbohydrates followed by elimination to afford unsaturated, derivatives. Solubility of amylose and amylopectin in DMSO has been evaluated.

Application

DMSO has been used to prepare the DMSO-phosphate buffer solution, which was used to dissolve the selective ETB (ET = endothelin) receptor antagonist in a study. It has been used for the preparation of nysatin solution.
Dimethyl sulfoxide may be used in the synthesis of the following:
  • α, β−Unsaturated carbonyl sugar derivatives.
  • Conversion of secondary bromides or olefins to bromohydrin.
  • Synthesis of α-hydroxycarbonyls from corresponding ketones.

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytotoxicity of dimethyl sulfoxide (DMSO) in direct contact with odontoblast-like cells.
Hebling J, et al.
Dental Materials : Official Publication of the Academy of Dental Materials, 31(4), 399-405 (2015)
Synthesis of α, β-unsaturated, carbonyl sugar derivatives by methyl sulfoxide oxidation and elimination.
Mackie DM and Perlin AS.
Carbohydrate Research, 24(1), 67-85 (1972)
From simple organobromides or olefins to highly value-added bromohydrins: a versatile performance of dimethyl sulfoxide.
Song S, et al.
Green Chemistry, 17(5), 2727-2731 (2015)
Solubility behavior of granular corn starches in methyl sulfoxide (DMSO) as measured by high performance size exclusion chromatography.
Jackson DS.
Starch/Staerke, 43(11), 422-427 (1991)
David Jarriault et al.
Journal of visualized experiments : JoVE, (101)(101), e52652-e52652 (2015-08-15)
Analyzing the physiological responses of olfactory sensory neurons (OSN) when stimulated with specific ligands is critical to understand the basis of olfactory-driven behaviors and their modulation. These coding properties depend heavily on the initial interaction between odor molecules and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service