Skip to Content
Merck
Dimethyl sulfoxide European Pharmacopoeia (EP) Reference Standard

Dimethyl sulfoxide

Dimethyl sulfoxide European Pharmacopoeia (EP) Reference Standard
Synonym(s):
DMSO, Dimethyl sulfoxide
Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13

Products

Product Category
Brand
Boiling Point (°C)
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-20 of 30
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2