L511692
2:1 2,2′-Bipyridine:Cyclopentadienyl(n6-napthalene)ruthenium(II) hexafluorophosphate
AldrichCPR
Synonym(s):
Herzon Hydration Catalyst
About This Item
Recommended Products
SMILES string
F[P-](F)(F)(F)(F)F.C1(C2=NC=CC=C2)=CC=CC=N1.C3(C4=NC=CC=C4)=CC=CC=N3.[C]56[C][C][C][C][C]5C=CC=C6.[Ru+].[C]7[C][C][C][C]7
InChI
1S/2C10H8N2.C10H8.C5H5.F6P.Ru/c2*1-3-7-11-9(5-1)10-6-2-4-8-12-10;1-2-6-10-8-4-3-7-9(10)5-1;1-2-4-5-3-1;1-7(2,3,4,5)6;/h2*1-8H;1-8H;1-5H;;/q;;;;-1;+1
InChI key
YFGQTOCRCOVSBH-UHFFFAOYSA-N
Related Categories
Application
Temporal separation of catalytic activities allows anti-Markovnikov reductive functionalization of terminal alkynes
Other Notes
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
The Herzon group has developed highly active catalysts for the anti-Markovnikov hydration and reductive hydration of terminal alkynes to form aldehydes and linear alcohols, respectively.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service