Skip to Content
Merck
All Photos(2)

Key Documents

805009

Sigma-Aldrich

CX200

Umicore

Synonym(s):

(Pd tBu-Ind Cl)2, Hazari Pd catalyst dimer, Palladium(p-tert-butylindenyl) chloride dimer, [(η3-1-tert-Butylindenyl)(μ-Cl)Pd]2

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$455.00

$455.00


Estimated to ship onApril 10, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$455.00

About This Item

Empirical Formula (Hill Notation):
C26H30Cl2Pd2
CAS Number:
Molecular Weight:
626.26
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

$455.00


Estimated to ship onApril 10, 2025


Request a Bulk Order

Quality Level

Assay

95%

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

166 °C

SMILES string

CC(C)(C)[C@H]1[C@H]2[C@H](CCCC2)CC1.C[Pd]Cl.C[Pd]Cl.CC(C)(C)c3ccc4c3cccC4

Application

The following palladium dimer is a precatalyst scaffold for cross-coupling reactions originally reported by the Hazari research group. When used with an selected ligand, the precatalyst exhibits superior activity over a wide array of cross-coupling reactions and under a variety of conditions.

Legal Information

Product of Umicore

Product License
This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

Additional information available at www.pmc.umicore.com

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melvin P R, et al.
ACS Catalysis, 5, 3680-3680 (2015)

Articles

The Hazari group has developed an improved palladium precatalyst scaffold for a wide range of cross-coupling reactions

Related Content

Hazari group develops versatile Ni and Pd precatalysts for cross-coupling reactions, aiming for efficient conversion under mild conditions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service