Merck
  • Design and synthesis of conjugated azo-hydrazone analogues using nano BF
Design and synthesis of conjugated azo-hydrazone analogues using nano BF
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2017-09-02)
Zabiulla, V Vigneshwaran, A Begum Bushra, G S Pavankumar, B T Prabhakar, Shaukath Ara Khanum
ABSTRACT
Disrupted redox balance is implicated in multiple pathologies including malignant progression and tumor angiogenesis. In this investigation, we report the design and development of novel and effective ROS detoxifying azo-hydrazone molecules targeting malignant pathologies and neoangiogenesis. A series of azo-derivatives conjugated to hydrazones moieties (9a-j) were synthesized using Nano BF
Product Description
Supelco
2-Fluoroaniline, ≥99%
Sigma-Aldrich
p-Anisidine, 99%
Sigma-Aldrich
2-Methyl-8-quinolinol, 98%
Sigma-Aldrich
5-Chloro-2-nitrobenzaldehyde, technical grade
Sigma-Aldrich
2-Hydroxy-4-methoxybenzaldehyde, 98%
Sigma-Aldrich
2-Bromoaniline, 98%