Merck
  • Structure-efficiency relationships of zwitterionic detergents as protein solubilizers in two-dimensional electrophoresis.
Structure-efficiency relationships of zwitterionic detergents as protein solubilizers in two-dimensional electrophoresis.
Proteomics (2003-02-26)
Christophe Tastet, Stéphane Charmont, Mireille Chevallet, Sylvie Luche, Thierry Rabilloud
ABSTRACT
Several zwitterionic detergents differing in their polar heads, linker parts and hydrophobia tail were synthesized and evaluated for their efficiency in protein solubilizers for two-dimensional electrophoresis. A model system consisting of human red blood cell ghosts was used for this purpose. This study leads to the description of several new efficient detergents and allowed us to derive structural constraints for the design and synthesis of efficient detergents for two-dimensional electrophoresis. These constraints apply to the hydrophilic head (sulfobetaine but not carboxybetaine), to the hydrophobic tail (12 to 16 alkyl carbons long, linear alkyl or alkylaryl) and to the presence and nature of the linker between the hydrophilic head and hydrophobic tail.
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
Sigma-Aldrich
3-(N,N-Dimethylmyristylammonio)propanesulfonate, ≥98.0% (TLC)
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
3-(N,N-Dimethylmyristylammonio)propanesulfonate, ≥99% (TLC)
Sigma-Aldrich
3-(N,N-Dimethylmyristylammonio)propanesulfonate, ≥99% (TLC), BioXtra
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1