Merck
Inhibition of cholinergic neurotransmission by β
American journal of physiology. Renal physiology (2017-04-28)
Isabel Silva, Ana Filipa Costa, Sílvia Moreira, Fátima Ferreirinha, Maria Teresa Magalhães-Cardoso, Isabel Calejo, Miguel Silva-Ramos, Paulo Correia-de-Sá
ABSTRACT
The direct detrusor relaxant effect of β
Sigma-Aldrich
Hemicholinium-3, solid, ≥95% (HPLC)
Sigma-Aldrich
S-(4-Nitrobenzyl)-6-thioinosine, ≥98%, solid
Sigma-Aldrich
8-Cyclopentyl-1,3-dipropylxanthine, solid
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Monoclonal Anti-Vacht antibody produced in mouse, clone S6-38, purified immunoglobulin
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)