Skip to Content
Merck
All Photos(1)

Key Documents

G5251

Sigma-Aldrich

DL-Glyceraldehyde 3-phosphate solution

45-55 mg/mL in H2O

Synonym(s):

(2-Hydroxy-3-oxopropoxy)phosphonic acid, 3-Phosphoglyceraldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7O6P
CAS Number:
Molecular Weight:
170.06
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

form

liquid

concentration

45-55 mg/mL in H2O

color

colorless

cation traces

Ba: ≤100 μg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC(COP(O)(O)=O)C=O

InChI

1S/C3H7O6P/c4-1-3(5)2-9-10(6,7)8/h1,3,5H,2H2,(H2,6,7,8)

InChI key

LXJXRIRHZLFYRP-UHFFFAOYSA-N

Application

The level of Glyceraldehyde 3-phosphate dehydrogenase (GAPDH) activity in HEK 293 cells was aβeβed by measuring the reduction of NAD+ by DL-Glyceraldehyde 3-phosphate.[1]

Biochem/physiol Actions

Glyceraldehyde 3-phosphate is an intermediate in several metabolic pathways, including glycolysis and gluconeogenesis. It is bactericidal and a potent inhibitor of growth in E. coli.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wanxia Yao et al.
Cellular signalling, 62, 109328-109328 (2019-05-31)
Accumulating evidence indicates that long non-coding RNAs (lncRNAs) play an important role in autism. Herein, we delineated the functions of LOC101927196 and its potential mitigation effect on a rat model of autism. We retrieved various bioinformatics databases and websites to
J Li et al.
European review for medical and pharmacological sciences, 23(14), 6091-6104 (2019-08-01)
The previous research revealed that long noncoding RNAs (lncRNAs) play a vital role in the development of hepatocellular carcinoma (HCC). To further discuss the underlying mechanisms of lncRNA DCST1-AS1 in the pathogenesis of HCC. We screened the abnormally expressed genes
Mohammed Nimir et al.
Cells, 8(7) (2019-07-11)
Detection of androgen receptor (AR) variant 7 (AR-V7) is emerging as a clinically important biomarker in castrate resistant prostate cancer (CRPC). Detection is possible from tumor tissue, which is often inaccessible in the advanced disease setting. With recent progress in
Bo Jia et al.
Aging, 11(14), 4990-5007 (2019-07-25)
Tongue squamous cell carcinoma (TSCC), the most common epithelial cancer identified in the oral cavity, has become one of the most common malignancies across the developing countries. Increasing evidence indicates that long non-coding RNAs (lncRNAs) serve as important regulators in
Noriyuki Akahoshi et al.
International journal of molecular sciences, 20(14) (2019-07-20)
Elevated plasma homocysteine levels are considered as a risk factor for cardiovascular diseases as well as preeclampsia-a pregnancy disorder characterized by hypertension and proteinuria. We previously generated mice lacking cystathionine γ-lyase (Cth) as cystathioninuria models and found them to be

Articles

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

See All

Questions

1–3 of 3 Questions  
  1. Does 25MG mean 25 MG of of 45-55 mg/ml in H2O

    1 answer
    1. Yes, this is correct. The lot specific concentration is reported in the product Certificate of Analysis. Please see the link below to review a sample or lot specific Certificate:
      https://www.sigmaaldrich.com/product/sigma/g5251#product=documentation

      Helpful?

  2. 45-55 mg/mL in H2O 일때 정확한 몰농도를 알 수 있나요?

    1 answer
    1. The molecular weight of the Glyceraldehyde 3-phosphate is 170.06 g/mol. The listed concentration of 45-55mg/ml is equivalent to 45-50g/L. This product will have a range of approximately 0.264-0.323M. The concentration is lot-specific and is reported on the Certificate of Analysis.

      Helpful?

  3. 45-55mg/ml 라고 표기되어 있는데, 정확한 몰농도를 알 수 있을까요?

    1 answer
    1. The exact concentration will be listed on the lot specific Certificate of Analysis. Please navigate to the DOCUMENTATION section of the Product Detail Page to access a Certificate under ‘Certificate of Analysis’:
      https://www.sigmaaldrich.com/product/sigma/g5251#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service