Merck
  • Inhibition effects of perfluoroalkyl acids on progesterone production in mLTC-1.
Inhibition effects of perfluoroalkyl acids on progesterone production in mLTC-1.
Journal of environmental sciences (China) (2017-06-03)
Wei Zhao, Ruina Cui, Jianshe Wang, Jiayin Dai
ABSTRACT
Perfluoroalkyl substances (PFASs) are a class of fluorine substituted carboxylic acid, sulfonic acid and alcohol, structurally similar to their corresponding parent compounds. Previous study demonstrated the potential endocrine disruption and reproductive toxicity of perfluorooctane sulfonic acid and perfluorooctanoic acid, two dominant PFASs in animals and humans. We explored the relationship between eleven perfluoroalkyl acids (PFAAs) with different carbon chain length and their ability to inhibit progesterone production in mouse Leydig tumor cells (mLTC-1). We found an obvious dose-response relationship between progesterone inhibition rate and PFAA exposure concentration in mLTC-1. The relative inhibition rate of progesterone by PFAAs was linearly related to the carbon chain length and molar refractivity of PFAAs. Mitochondrial membrane potential (MMP) decreased after PFAA exposure at the half-maximal inhibitory effect concentration (IC
Sigma-Aldrich
Perfluorodecanoic acid, 98%
Sigma-Aldrich
Tricosafluorododecanoic acid, 95%
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Perfluorotetradecanoic acid, 96%