Merck
  • MiR-129-5p Inhibits Proliferation and Invasion of Chondrosarcoma Cells by Regulating SOX4/Wnt/β-Catenin Signaling Pathway.
MiR-129-5p Inhibits Proliferation and Invasion of Chondrosarcoma Cells by Regulating SOX4/Wnt/β-Catenin Signaling Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2017-05-24)
Peng Zhang, Jifeng Li, Yuze Song, Xiao Wang
ABSTRACT
Recently, microRNAs (miRNA) have been identified as novel regulators in Chondrosarcoma (CHS). This study was aimed to identify the roles of miR-129-5p-5p in regulation of SOX4 and Wnt/β-catenin signaling pathway, as well as cell proliferation and apoptosis in chondrosarcomas. Tissue samples were obtained from chondrosarcoma patients. Immunohistochemistry, real-time quantitative RT-PCR (RT-qPCR) and western blot analysis were performed to detect the expressions of miR-129-5p and SOX4. Luciferase assay was conducted to confirm that miR-129-5p directly targeted SOX4 mRNA. Manipulations of miR-129-5p and SOX4 expression were achieved through cell transfection. Cell proliferation, migration and apoptosis were evaluated by CCK-8 assay, colony forming assay, wound healing assay and flow cytometry in vitro. For in vivo experiment, the tumor xenograft model was established to evaluate the effects of miR-129-5p and SOX4 on chondrosarcomas. The expression of miR-129-5p was significantly down-regulated in chondrosarcoma tissues as well as cells in comparison with normal ones, while SOX4 was over-activated. Further studies suggested that miR-129-5p suppressed cell proliferation, migration and promoted apoptosis by inhibiting SOX4 and Wnt/β-catenin pathway. MiR-129-5p inhibits the Wnt/β-catenin signaling pathway by targeting SOX4 and further suppresses cell proliferation, migration and promotes apoptosis in chondrosarcomas.
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
NuCLEAR Extraction Kit, For mammalian tissue or cultured cells
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Roche
Anti-c-myc, from mouse IgG1κ
Sigma-Aldrich
Anti-SOX4 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Cyclin A antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Mcl-1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
QuantiPro BCA Assay Kit, for 0.5-30 μg/ml protein
Sigma-Aldrich
Anti-Histone H3 (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-β-Catenin antibody produced in rabbit, whole antiserum