Merck
Differential Contribution of Ca
Frontiers in cellular neuroscience (2017-07-18)
Eric C Lin, Crescent L Combe, Sonia Gasparini
ABSTRACT
Temporal lobe epilepsy is characterized by recurrent seizures in one or both temporal lobes of the brain; some
Sigma-Aldrich
9-Phenanthrol, technical grade