Merck
  • Heart failure modulates electropharmacological characteristics of sinoatrial nodes.
Heart failure modulates electropharmacological characteristics of sinoatrial nodes.
Experimental and therapeutic medicine (2017-03-30)
Shih-Lin Chang, Hui-Lun Chuang, Yao-Chang Chen, Yu-Hsun Kao, Yung-Kuo Lin, Yung-Hsin Yeh, Shih-Ann Chen, Yi-Jen Chen
ABSTRACT
The impact of heart failure (HF) on sinoatrial node (SAN) channel regulation and electropharmacological responses has remained elusive. The present study aimed to investigate the effects of HF on the electrical activity of SANs with and without pharmacological interventions. Action potentials (APs) were recorded in isolated SANs from normal rabbits (control) and those with HF (rapid ventricular pacing for 4 weeks) prior to and after administration of a funny current blocker (ivabradine; 0.1, 0.3, 3 or 10 µM), a calmodulin kinase II inhibitor (KN-93; 0.3 or 3 µM), a sarcoplasmic reticulum Ca2+ release inhibitor (ryanodine; 0.3 or 3 µM), a sodium current inhibitor (tetrodotoxin; 1, 3 or 10 µM) and a late sodium current inhibitor (ranolazine; 10 µM). Western blot analysis was used to investigate the protein expression in SANs from normal rabbits and those with HF. Control SANs had a higher beating rate than SANs from rabbits with HF (2.3±0.1 vs. 1.5±0.1 Hz; P<0.001). Similarly, ivabradine (10 µM), KN-93 (3 µM), ranolazine (10 µM) and ryanodine (3 µM) decreased the beating rates of SANs in the control (n=6) and HF (n=6) groups. Ivabradine treatment resulted in a higher incidence of AP block in HF vs. control SANs (66.7 vs. 0%; P<0.05). Tetrodotoxin (1, 3 or 10 µM) decreased the beating rate to a higher extent in SANs from rabbits with HF than in those from control rabbits and induced a higher incidence of AP block (66.7 vs. 0%; P<0.05). Furthermore, SANs from rabbits with HF had higher protein levels of phospholamban (PLB) and lower levels of hyperpolarization-activated cyclic nucleotide-gated potassium channel 4, ryanodine receptor and phosphorylated PLB than control SANs. In conclusion, HF modulates electropharmacological responses in the SAN by channel regulation, which may result in SAN dysfunction.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Peroxidase Conjugated, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Anti-HCN4 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-Actin (α-Sarcomeric) antibody produced in mouse, clone 5C5, ascites fluid
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Peroxidase Conjugated, H+L, 1 mg/mL (after reconstitution), Chemicon®