Merck
  • CTCF cooperates with noncoding RNA MYCNOS to promote neuroblastoma progression through facilitating MYCN expression.
CTCF cooperates with noncoding RNA MYCNOS to promote neuroblastoma progression through facilitating MYCN expression.
Oncogene (2015-11-10)
X Zhao, D Li, J Pu, H Mei, D Yang, X Xiang, H Qu, K Huang, L Zheng, Q Tong
ABSTRACT
Previous studies have indicated the important roles of MYCN in tumorigenesis and progression of neuroblastoma (NB), the most common extracranial solid tumor derived from neural crest in childhood. However, the regulatory mechanisms of MYCN expression in NB still remain largely unknown. In this study, through mining public microarray databases and analyzing the cis-regulatory elements and chromatin immunoprecipitation data sets, we identified CCCTC-binding factor (CTCF) as a crucial transcription factor facilitating the MYCN expression in NB. RNA immunoprecipitation, RNA electrophoretic mobility shift assay, RNA pull down and in vitro binding assay indicated the physical interaction between CTCF and MYCN opposite strand (MYCNOS), a natural noncoding RNA surrounding the MYNC promoter. Gain- and loss-of-function studies revealed that MYCNOS facilitated the recruitment of CTCF to its binding sites within the MYCN promoter to induce chromatin remodeling, resulting in enhanced MYCN levels and altered downstream gene expression, in cultured NB cell lines. CTCF cooperated with MYCNOS to suppress the differentiation and promote the growth, invasion and metastasis of NB cells in vitro and in vivo. In clinical NB tissues and cell lines, CTCF and MYCNOS were upregulated and positively correlated with MYCN expression. CTCF was an independent prognostic factor for unfavorable outcome of NB, and patients with high MYCNOS expression had lower survival probability. Taken together, these results demonstrate that CTCF cooperates with noncoding RNA MYCNOS to exhibit oncogenic activity that affects the aggressiveness and progression of NB through transcriptional upregulation of MYCN.
Sigma-Aldrich
ChIPAb+ Trimethyl-Histone H3 (Lys27) - ChIP Validated Antibody and Primer Set, from rabbit, purified by using Protein A
Sigma-Aldrich
Anti-CTCF Antibody, serum, Upstate®
Sigma-Aldrich
ChIPAb+ RNA Pol II - ChIP Validated Antibody and Primer Set, from mouse
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys4) Antibody, clone AW30, rabbit monoclonal, culture supernatant, clone AW30, Upstate®
Sigma-Aldrich
Magna RIP® RNA-Binding Protein Immunoprecipitation Kit, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.