Skip to Content
Merck
All Photos(2)

Key Documents

17-622

Sigma-Aldrich

ChIPAb+ Trimethyl-Histone H3 (Lys27) - ChIP Validated Antibody and Primer Set

from rabbit, purified by using Protein A

Synonym(s):

Chip Antibody and primer set, H3K27me3 ChIP, H3K27me3, Histone H3 (tri methyl K27), Histone H3K27me3, Histone H3K27me3 ChIP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.32

biological source

rabbit

Quality Level

clone

polyclonal

purified by

using Protein A

species reactivity

human, mouse

manufacturer/tradename

ChIPAb+
Upstate®

technique(s)

ChIP: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... H3F3B(3021)

General description

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot,
every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context.
Each set also includes a negative control antibody to ensure specificity of the ChIP reaction. The ChIPAb+ Trimethyl-Histone H3 (Lys27) set includes the Anti-Trimethyl-Histone H3 (Lys27) antibody, a negative control antibody (purified rabbit IgG), and qPCR primers which amplify a 139 bp region of human alpha-Satellite. The Trimethyl-Histone H3 (Lys27) and negative control antibodies are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of Trimethyl-Histone H3 (Lys27)-associated chromatin.
The methylation of histones can occur on two different residues: arginine or lysine. Histone methylation can be associated with transcriptional activation or repression, depending on the methylated residue. Lysine 27 of histone H3 can be mono-, di- or trimethylated (Histone H3 monomethyl Lys27, Histone H3 dimethyl Lys27 or Histone H3 trimethyl Lys27) by different histone methyltransferases such as EZH2 or NSD3. Methylation of this residue is mainly associated with transcriptional repression.

Specificity

Broad species cross-reactivity expected.
trimethyl-Histone H3 (Lys27)

Immunogen

Epitope: Trimethyl Lys27
KLH-conjugated, synthetic 2X-branched peptide containing the sequence ...AR(me3K)SAP... in which me3K corresponds to trimethyl-lysine at residue 27 of human Histone H3.

Application

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from 2x106 HeLa cells were subjected to chromatin immunoprecipitation using 4 μg purified anti-trimethyl-Histone H3 (Lys27) antibody or normal rabbit IgG and the Magna ChIP A kit (Cat. #17-610). Successful enrichment of trimethyl-Histone H3 (Lys27) associated DNA fragments was verified by qPCR using ChIP Primers GAPDH flanking the human GAPDH promoter and primers targeting the promoter of human MyoD.
Please refer to the EZ-Magna ChIP A (Cat. #17-408) or EZChIP (Cat. #17-371) kit protocols for experimental details.

Western Blot Analysis:
Recombinant Histone H3 (Lane 2) and HeLa cell acid extracts (Lane 1) were resolved by electrophoresis, transferred to nitrocellulose and probed with a 1:5000 dilution of anti- trimethyl histone H3 Lys27 (0.4 μg/mL).
Proteins were visualized using a goat anti-rabbit secondary antibody conjugated to HRP and a chemiluminescence detection system (Please see figures).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Trimethyl-Histone H3 (Lys27) ChIP validated antibody & primer set including the ChIP-grade antibody & the specific control PCR primers used for chromatin immunoprecipitation of H3K27Me3.

Packaging

25 assays per kit, ~4μL per chromatin immunoprecipitation.

Components

Anti-trimethyl Histone H3(Lys27) polyclonal, 1 vial

Polyclonal control rabbit serum, 1 vial

Human alpha-Satellite primer set, 1 vial

Quality

Chromatin Immunoprecipitation:
Sonicated Chromatin prepared from 3x106 NIH3T3 L1 cells were subjected to chromatin immunoprecipitation using 4 μg purified antibody or normal rabbit IgG and the Magna ChIP® A kit (Cat. #17-610). Successful enrichment of trimethyl-Histone H3
(Lys27) associated DNA fragments was verified by qPCR using ChIP Primers human alpha-Satellite (Please see figures).
Please refer to the EZ-Magna ChIP A (Cat. #17-408) or EZ-ChIP (Cat. #17-371) kit protocols for experimental details.

Target description

17kDa

Physical form

Anti-trimethyl-Histone H3 (Lys27) (rabbit polyclonal IgG). One vial containing 100 μg protein A purified IgG in 100 μL buffer containing 0.1 M Tris-Glycine, pH 7.4, 0.15M NaCl with 0.05% sodium azide. Store at -20°C.

Normal Rabbit IgG. One vial containing 125 μg purified Rabbit IgG in 125 μL storage buffer containing 0.1% sodium azide. Store at -20°C.

ChIP Primers human alpha-Satellite. One vial containing 75 μL of 5 μM of each control primer specific for human alpha-Satellite. Store at -20°C.
FOR: CTG CAC TAC CTG AAG AGG AC
REV: GAT GGT TCA ACA CTC TTA CA

Storage and Stability

1 year at -20°C from date of shipment

Analysis Note

Control
1 vial containing 75μl of 5μM of each control primer specific for human alpha-Satellite. Store at -20°C.

FOR: CTG CAC TAC CTG AAG AGG AC

REV: GAT GGT TCA ACA CTC TTA CA

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-wide bovine H3K27me3 modifications and the regulatory effects on genes expressions in peripheral blood lymphocytes.
He, Y; Yu, Y; Zhang, Y; Song, J; Mitra, A; Zhang, Y; Wang, Y; Sun, D; Zhang, S
Testing null
Transgene- and locus-dependent imprinting reveals allele-specific chromosome conformations.
Lonfat, N; Montavon, T; Jebb, D; Tschopp, P; Nguyen Huynh, TH; Zakany, J; Duboule, D
Proceedings of the National Academy of Sciences of the USA null
Judit Balog et al.
Epigenetics, 10(12), 1133-1142 (2015-11-18)
Facioscapulohumeral muscular dystrophy is caused by incomplete epigenetic repression of the transcription factor DUX4 in skeletal muscle. A copy of DUX4 is located within each unit of the D4Z4 macrosatellite repeat array and its derepression in somatic cells is caused
Claudio D'Addario et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 25(3), 1069-1075 (2010-11-26)
Ethanol alters neural activity through interaction with multiple neurotransmitters and neuromodulators. The endogenous opioid system seems to play a key role, since the opioid receptor antagonist naltrexone (ReVia®) attenuates craving for alcohol. We recently reported that ethanol and acetaldehyde, the
Modular utilization of distal cis-regulatory elements controls Ifng gene expression in T cells activated by distinct stimuli.
Balasubramani, A; Shibata, Y; Crawford, GE; Baldwin, AS; Hatton, RD; Weaver, CT
Immunity null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service