Merck
  • Hydrophobicity and antioxidant activity acting together for the beneficial health properties of nordihydroguaiaretic acid.
Hydrophobicity and antioxidant activity acting together for the beneficial health properties of nordihydroguaiaretic acid.
Food & function (2015-05-01)
Luana Chiquetto Paracatu, Carolina Maria Quinello Gomes de Faria, Maria Luiza Zeraik, Camila Quinello, Camila Rennó, Patrícia Palmeira, Luiz Marcos da Fonseca, Valdecir Farias Ximenes
ABSTRACT
Nordihydroguaiaretic acid (NDGA) and rosmarinic acid (RA), phenolic compounds found in various plants and functional foods, have known antioxidant and anti-inflammatory properties. In the present study, we comparatively investigated the importance of hydrophobicity and oxidisability of NDGA and RA, regarding their antioxidant and pharmacological activities. Using a panel of cell-free antioxidant protocols, including electrochemical measurements, we demonstrated that the anti-radical capacities of RA and NDGA were similar. However, the relative capacity of NDGA as an inhibitor of NADPH oxidase (ex vivo assays) was significantly higher compared to RA. The inhibitory effect on NADPH oxidase was not related to simple scavengers of superoxide anions, as confirmed by oxygen consumption by the activated neutrophils. The higher hydrophobicity of NDGA was also a determinant for the higher efficacy of NDGA regarding the inhibition of the release of hypochlorous acid by PMA-activated neutrophil and cytokine (TNF-α and IL-10) production by Staphylococcus aureus-stimulated peripheral blood mononuclear cells. In conclusion, although there have been extensive studies about the pharmacological properties of NDGA, our study showed, for the first time, the importance not only of its antioxidant activity, but also its hydrophobicity as a crucial factor for pharmacological action.
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
4′-Hydroxy-3′-methoxyacetophenone, 98%
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Acetovanillone, ≥98%, FG
Sigma-Aldrich
Rosmarinic acid, 96%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
Dihydrorhodamine 123, ≥95%
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
SAFC
Taurine
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%