Skip to Content
Merck
All Photos(1)

Key Documents

D1054

Sigma-Aldrich

Dihydrorhodamine 123

≥95% purity, powder

Synonym(s):

DHR 123; Methyl 2-(3,6-diamino-9H-xanthen-9-yl)benzoate; Benzoic acid, 2-(3,6-diamino-9H-xanthen-9-yl)-, methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18N2O3
CAS Number:
Molecular Weight:
346.38
Beilstein:
5989965
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Dihydrorhodamine 123, ≥95%

Quality Level

Assay

≥95%

form

powder

technique(s)

titration: suitable

color

pink

solubility

DMSO: >5 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

COC(=O)c1ccccc1C2c3ccc(N)cc3Oc4cc(N)ccc24

InChI

1S/C21H18N2O3/c1-25-21(24)15-5-3-2-4-14(15)20-16-8-6-12(22)10-18(16)26-19-11-13(23)7-9-17(19)20/h2-11,20H,22-23H2,1H3

InChI key

FNEZBBILNYNQGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dihydrorhodamine 123 (DHR123) is an uncharged non-fluorescent dye and a derivative of rhodamine 123 (R123). Dihydrorhodamine 123 (DHR123) is oxidized to fluorescent R123 within cells in the presence of reactive oxygen species and it localizes in mitochondria. It is an analog of 2,7-Dichlorodihydrofluorescein (DCDHF), lipophilic and diffuses through cell membrane.

Application

Dihydrorhodamine 123 (DHR) has been used for the detection of free intracellular radicals or ROS (reactive oxygen species) in head kidney-derived macrophages. It has also been used in DHR assay in red blood cells and in oxidative burst assay in neutrophils.

Biochem/physiol Actions

Dihydrorhodamine 123 detects superoxide ions at the single-cell level and is used for probing neutrophils superoxide generation in a respiratory burst event. It is also used for measuring oxidative stress in macrophages and human spermatozoa.
Dihydrorhodamine 123 is used for the detection of ROS (reactive oxygen species). Presence of ROS oxidizes dihydrorhodamine 123 to the fluorescent derivative rhodamine 123. The reaction happens in the presence of a peroxidase or a similar catalyst.

Substrates

Peroxidase substrate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of the calcium-mediated response to alkaline stress in Saccharomyces cerevisiae.
Viladevall L, et al.
The Journal of Biological Chemistry, 279, 43614-43624 (2004)
Phosphatase Wip1 negatively regulates neutrophil development through p38 MAPK-STAT1
Liu G, et al.
Blood, 121(3), 519-529 (2013)
Dichlorodihydrofluorescein and dihydrorhodamine 123 are sensitive indicators of peroxynitrite in vitro: implications for intracellular measurement of reactive nitrogen and oxygen species
Crow JP
Nitric Oxide, 1(2), 145-157 (1997)
DHR123: an alternative probe for assessment of ROS in human spermatozoa
Kiani-Esfahani A, et al.
Systems Biology in Reproductive Medicine, 58(3), 168-174 (2012)
Dihydrorhodamine 123: a fluorescent probe for superoxide generation?
HENDERSON LM and CHAPPELL JB
European Journal of Biochemistry, 217(3), 973-980 (1993)

Articles

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service