Merck
  • Monodisperse AgSbS2 nanocrystals: size-control strategy, large-scale synthesis, and photoelectrochemistry.
Monodisperse AgSbS2 nanocrystals: size-control strategy, large-scale synthesis, and photoelectrochemistry.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-06-23)
Bin Zhou, Mingrun Li, Yihui Wu, Chi Yang, Wen-Hua Zhang, Can Li
ABSTRACT
We report an efficient approach to the synthesis of AgSbS2 nanocrystals (NCs) by colloidal chemistry. The size of the AgSbS2 NCs can be tuned from 5.3 to 58.3 nm with narrow size distributions by selection of appropriate precursors and fine control of the experimental conditions. Over 15 g of high-quality AgSbS2 NCs can be obtained from one single reaction, indicative of the up-scalability of the present synthesis. The resulting NCs display strong absorptions in the visible-to-NIR range and exceptional air stability. The photoelectrochemical measurements indicate that, although the pristine AgSbS2 NC electrodes generate a cathodic photocurrent with a relatively small photocurrent density and poor stability, both of them can be significantly improved subject to CdS surface modification, showing promise in solar energy conversion applications.
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Antimony(III) acetate, 99.99% trace metals basis
Sigma-Aldrich
Sulfur, 99.998% trace metals basis
Sigma-Aldrich
Hexamethyldisilathiane, synthesis grade
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
Sulfur, powder, 99.98% trace metals basis
Sigma-Aldrich
1-Dodecanethiol, ≥98%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
SAFC
Oleylamine, ≥98% (primary amine)
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Octanoic acid, ≥99%
Sigma-Aldrich
Dodecylamine, ≥99%
Sigma-Aldrich
Octanoic acid, ≥98%
Sigma-Aldrich
Octanoic acid, natural, ≥98%, FG
Sigma-Aldrich
Antimony(III) chloride, ≥99.95% trace metals basis
Sigma-Aldrich
Octanoic acid, ≥98%, FG
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Dodecylamine, 98%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%