Merck
  • Collision-induced dissociation of aminophospholipids (PE, MMPE, DMPE, PS): an apparently known fragmentation process revisited.
Collision-induced dissociation of aminophospholipids (PE, MMPE, DMPE, PS): an apparently known fragmentation process revisited.
Analytical and bioanalytical chemistry (2015-01-31)
Ernst Pittenauer, Pavel Rehulka, Wolfgang Winkler, Günter Allmaier
ABSTRACT
A new type of low-mass substituted 4-oxazolin product ions of [M + H](+) precursor ions of aminophospholipids (glycerophosphatidylethanolamine, glycerophosphatidyl-N-methylethanolamine, glycerophosphatidyl-N,N-dimethylethanolamine, glycerophosphatidylserine) resulting from high-energy collision-induced dissociation (matrix-assisted laser desorption/ionization time-of-flight/reflectron time-of-flight mass spectrometry) and low-energy collision-induced dissociation (e.g., electrospray ionization quadrupole reflectron time-of-flight mass spectrometry) with accurate mass determination is described; these were previously misidentified as CHO-containing radical cationic product ions. The mechanism for the formation of these ions is proposed to be via rapid loss of water followed by cyclization to an 11-membered-ring transition state for the sn-1 fatty acid substituent and to a ten-membered-ring transition state for the sn-2 fatty acid substituent, and via final loss of monoacylglycerol phosphate, leading to substituted 4-oxazolin product ions. The minimum structural requirement for this interesting skeletal rearrangement fragmentation is an amino group linked to at least one hydrogen atom (i.e., ethanolamine, N-methylethanolamine, serine). Therefore, N,N-dimethylethanolamine derivates do not exhibit this type of fragmentation. The analytical value of these product ions is given by the fact that by post source decay and particularly high-energy collision-induced dissociation achieved via matrix-assisted laser desorption/ionization time-of-flight/reflectron time-of-flight mass spectrometry, the sn-2-related substituted 4-oxazolin product ion is always significantly more abundant than the sn-1-related one, which is quite helpful for detailed structural analysis of complex lipids. All other important product ions found are described in detail (following our previously published glycerophospholipid product ion nomenclature; Pittenauer and Allmaier, Int. J. Mass. Spectrom. 301:90-1012, 2011).
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
1-Amino-2-methyl-2-propanol, 95% anhydrous basis
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
2-(Methylamino)ethanol, ≥98%
Sigma-Aldrich
2′,4′,6′-Trihydroxyacetophenone monohydrate, 98%
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))