Skip to Content
Merck
All Photos(2)

Key Documents

T64602

Sigma-Aldrich

2′,4′,6′-Trihydroxyacetophenone monohydrate

98%

Synonym(s):

2-Acetylphloroglucinol, THAP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)3C6H2COCH3·H2O
CAS Number:
Molecular Weight:
186.16
Beilstein:
9288656
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

219-221 °C (lit.)

SMILES string

O.CC(=O)c1c(O)cc(O)cc1O

InChI

1S/C8H8O4.H2O/c1-4(9)8-6(11)2-5(10)3-7(8)12;/h2-3,10-12H,1H3;1H2

InChI key

GDSIBPPJKSBCMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Tintoré et al.
International journal of molecular sciences, 16(11), 27625-27639 (2015-11-26)
Au- and iron-based magnetic nanoparticles (NPs) are promising NPs for biomedical applications due to their unique properties. The combination of a gold coating over a magnetic core puts together the benefits from adding the magnetic properties to the robust chemistry
Liang Xu et al.
Nucleic acid therapeutics, 25(4), 219-225 (2015-07-21)
DNA triplexes with hydrophobic modifications were designed and evaluated for their activity as inhibitors of the cell fusion of human immunodeficiency virus type 1 (HIV-1). Triplex inhibitors displayed low micromolar activities in the cell-cell fusion assay and nanomolar activities in
Chaitanya V Mungi et al.
Life (Basel, Switzerland), 5(1), 65-84 (2015-01-09)
Prebiotic polymerization had to be a nonenzymatic, chemically driven process. These processes would have been particularly favored in scenarios which push reaction regimes far from equilibrium. Dehydration-rehydration (DH-RH) cycles are one such regime thought to have been prevalent on prebiotic
Gerard Artigas et al.
The Journal of organic chemistry, 80(4), 2155-2164 (2015-01-21)
We describe the synthesis and characterization of ametantrone-containing RNA ligands based on the derivatization of this intercalator with two neamine moieties (Amt-Nea,Nea) or with one azaquinolone heterocycle and one neamine (Amt-Nea,Azq) as well as its combination with guanidinoneamine (Amt-NeaG4). Biophysical
David J Harvey
Mass spectrometry reviews, 34(3), 268-422 (2014-05-28)
This review is the sixth update of the original article published in 1999 on the application of MALDI mass spectrometry to the analysis of carbohydrates and glycoconjugates and brings coverage of the literature to the end of 2010. General aspects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service