Merck
  • Solvent bar micro-extraction: Improving hollow fiber liquid phase micro-extraction applicability in the determination of Ni in seawater samples.
Solvent bar micro-extraction: Improving hollow fiber liquid phase micro-extraction applicability in the determination of Ni in seawater samples.
Talanta (2015-05-25)
Juan J Pinto, Mabel Martín, Belén Herce-Sesa, José A López-López, Carlos Moreno
ABSTRACT
During the last decade, hollow fiber liquid phase micro-extraction (HF-LPME) has become an attractive alternative in sample treatment for the analysis of trace metals in seawater. If compared with other similar methodologies, its main advantages are associated to a higher stability of the organic solution contained into the pores of the fiber, which acts as a lipophilic membrane during the extraction process. However there are some remaining problems that makes its use difficult, mostly related to the need of increasing the rate of analysis and improving portability. In this paper a novel three phase solvent bar micro-extraction (3PSBME) for the fiber device has been proposed. Its main advantage is that the 3PSBME device can be left free in the sample. This way the system is portable, and no special support is needed leading to the possibility of simultaneous extraction of several samples. In this work, multivariate central composite design of experiment has been carried out to optimize Ni pre-concentration using di-2-ethylhexyl phosphoric acid (DEHPA) as extractant and HNO3 as acceptor agent. Factors influencing extraction have been the pH in the sample and the fiber length. For seawater samples, Ni can be pre-concentrated 11 times in 140 min. The method presents RSD 9.42% and limit of detection 44 ng L(-1), using GFAAS for instrumental determination. It has been applied for determination of Ni in seawater, including a reference material CRM-403 proving its applicability.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Diethyldithiocarbamic acid diethylammonium salt, 97%
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis