Merck
  • Reversed-phase Chromatography in an Extended Nanospace: Separating Amino Acids in Short and Long Nanochannels.
Reversed-phase Chromatography in an Extended Nanospace: Separating Amino Acids in Short and Long Nanochannels.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2015-11-13)
Adelina Smirnova, Hisashi Shimizu, Kazuma Mawatari, Takehiko Kitamori
ABSTRACT
Micro- and nanofluidics has attracted much attention, particularly concerning single-cell analysis when small amounts of liquids are examined. In present work we successfully fabricated extended-nano channels that were more narrow and shorter (2 mm) as well as wider and longer (10 mm), and accomplished a reversed-phase HPLC separation of labeled amino acids on these channels after octadecylsilylation (ODS). The separation performance characteristics were compared for both types of nano spaces. At an equal amount of pressure, the longer extended-nano channels showed permeability that was one-order higher (K = 47 × 10(-14) m(2)) and separation impedance (E = 13) that was one-order lower than that of the shorter version. Also, the separation plate number for the longer channel was 4000 with a plate height of 2.5 μm. Both channels have advantages for use in single-cell analysis. The longer channel can be applied for the separation of macromolecules (proteomics), while the short version is more applicable to small molecules (amino acids).
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium perchlorate hydrate, 99.99% trace metals basis
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
SAFC
L-Proline
SAFC
L-Valine
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
Chloro(dimethyl)octadecylsilane, 95%
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O