Merck
  • Catalytic and charge transfer properties of transition metal dichalcogenides arising from electrochemical pretreatment.
Catalytic and charge transfer properties of transition metal dichalcogenides arising from electrochemical pretreatment.
ACS nano (2015-04-22)
Xinyi Chia, Adriano Ambrosi, Zdenek Sofer, Jan Luxa, Martin Pumera
ABSTRACT
Layered transition metal dichalcogenides (TMDs) have been the center of attention in the scientific community due to their properties that can be tapped on for applications in electrochemistry and hydrogen evolution reaction (HER) catalysis. We report on the effect of electrochemical treatment of exfoliated MoS2, WS2, MoSe2 and WSe2 nanosheets toward the goal of activating the electrochemical and HER catalytic properties of the TMDs. In particular, electrochemical activation of the heterogeneous electron transfer (HET) abilities of MoS2, MoSe2 and WSe2 is achieved via reductive treatments at identified reductive potentials based on their respective inherent electrochemistry. Comparing all TMDs, the charge transfer activation is most accentuated in MoSe2 and can be concluded that Mo metal and Se chalcogen type are more susceptible to electrochemical activation than W metal and S chalcogen type. With regards to the HER, we show that while MoS2 displayed enhanced performance when subjected to electrochemical reduction, WS2 fared worse upon oxidation. On the other hand, the HER performance of MoSe2 and WSe2 is independent of electrochemical redox treatment. We can conclude therefore that for the HER, S-containing TMDs are more responsive to redox treatment than compounds with the Se chalcogen. Our findings are beneficial toward understanding the electrochemistry of TMDs and the extent to which activation by electrochemical means is effective. In turn, when such knowledge is administered aptly, it will be promising for electrochemical uses.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
tert-Butyllithium solution, 1.7 M in pentane
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
tert-Butyllithium solution, 1.6-3.2 M in heptane
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
SAFC
Sodium chloride solution, 5 M