Merck
  • Allocation dynamics of recently fixed carbon in beech saplings in response to increased temperatures and drought.
Allocation dynamics of recently fixed carbon in beech saplings in response to increased temperatures and drought.
Tree physiology (2015-04-17)
Carola H Blessing, Roland A Werner, Rolf Siegwolf, Nina Buchmann
ABSTRACT
The response of carbon allocation to drought has often been studied in terms of short-term transport velocity of recently fixed carbon from leaves to roots and root respiration. However, its dynamic response to other environmental conditions, e.g., to changes in temperature, is less clear. Here, we investigated the effects of drought, increased temperatures and their combination on transport velocity as well as on distribution of recent photoassimilates for different compounds, such as sugars, starch, organic acids and amino acids. We used a (13)CO(2) pulse-labelling approach and studied the recovery of (13)C in different plant tissues and compounds of beech saplings (Fagus sylvatica L.) during a 9-day chase period. Neither total dry biomass nor dry weights of leaves or roots were affected by drought or increased temperatures. Generally, the fast transfer of recently fixed assimilates from leaves to roots took about 1 day, while (13)C enrichment in soil CO(2) efflux peaked only 2 days after labelling. Increased temperatures prolonged mean transfer times of recent photoassimilates from the leaves to the roots, probably caused by enhanced intermediate storage alongside basipetal transfer, clearly impacting short-term carbon allocation. This temperature effect was seen in the delayed peak in (13)C excess of root sugars, decoupling the roots from the leaves in the short term. On average, ∼40% of the (13)C label initially present in the plant was recovered in the roots (over all treatment combinations), providing strong evidence for preferred carbon allocation into the roots at the end of the growing season. Root starch was the principal compound for long-term storage of carbon, whereas leaf (transitory) starch was remobilized again after some days, exhibiting the longest mean residence times under dry and warm conditions. These observation clearly point to different functionalities of the same compound (i.e., starch) in different plant tissues and the crucial role of roots for long-term carbon storage.
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis