Merck
  • Shedding of syndecan-4 promotes immune cell recruitment and mitigates cardiac dysfunction after lipopolysaccharide challenge in mice.
Shedding of syndecan-4 promotes immune cell recruitment and mitigates cardiac dysfunction after lipopolysaccharide challenge in mice.
Journal of molecular and cellular cardiology (2015-10-10)
Mari E Strand, Jan Magnus Aronsen, Bjørn Braathen, Ivar Sjaastad, Heidi Kvaløy, Theis Tønnessen, Geir Christensen, Ida G Lunde
ABSTRACT
Inflammation is central to heart failure progression. Innate immune signaling increases expression of the transmembrane proteoglycan syndecan-4 in cardiac myocytes and fibroblasts, followed by shedding of its ectodomain. Circulating shed syndecan-4 is increased in heart failure patients, however the pathophysiological and molecular consequences associated with syndecan-4 shedding remain poorly understood. Here we used lipopolysaccharide (LPS) challenge to investigate the effects of syndecan-4 shedding in the heart. Wild-type mice (10mg/kg, 9h) and cultured neonatal rat cardiomyocytes and fibroblasts were subjected to LPS challenge. LPS increased cardiac syndecan-4 mRNA without altering full-length protein. Elevated levels of shedding fragments in the myocardium and blood from the heart confirmed syndecan-4 shedding in vivo. A parallel upregulation of ADAMTS1, ADAMTS4 and MMP9 mRNA suggested these shedding enzymes to be involved. Echocardiography revealed reduced ejection fraction, diastolic tissue velocity and prolonged QRS duration in mice unable to shed syndecan-4 (syndecan-4 KO) after LPS challenge. In line with syndecan-4 shedding promoting immune cell recruitment, expression of immune cell markers (CD8, CD11a, F4/80) and adhesion receptors (Icam1, Vcam1) were attenuated in syndecan-4 KO hearts after LPS. Cardiomyocytes and fibroblasts exposed to shed heparan sulfate-substituted syndecan-4 ectodomains showed increased Icam1, Vcam1, TNFα and IL-1β expression and NF-κB-activation, suggesting direct regulation of immune cell recruitment pathways. In cardiac fibroblasts, shed ectodomains regulated expression of extracellular matrix constituents associated with collagen synthesis, cross-linking and turnover. Higher syndecan-4 levels in the coronary sinus vs. the radial artery of open heart surgery patients suggested that syndecan-4 is shed from the human heart. Our data demonstrate that shedding of syndecan-4 ectodomains is part of the cardiac innate immune response, promoting immune cell recruitment, extracellular matrix remodeling and mitigating cardiac dysfunction in response to LPS.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
NF-κB Activation Inhibitor III, The NF-κB Activation Inhibitor III, also referenced under CAS 380623-76-7, controls the biological activity of NF-κB. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))