Merck
  • 1-Aza-15-Crown-5 Functionalized Graphene Oxide for 2D Graphene-Based Li⁺-ion Conductor.
1-Aza-15-Crown-5 Functionalized Graphene Oxide for 2D Graphene-Based Li⁺-ion Conductor.
Small (Weinheim an der Bergstrasse, Germany) (2015-03-12)
Moutusi Banerjee, Abhisek Gupta, Shyamal K Saha, Dipankar Chakravorty
ABSTRACT
Attachment of Li(+) ion on graphene surface to realize Li(+)-ion conductor is a real challenge because of the weak interaction between the ions and the functional groups of graphene oxide; although, a large number of theoretical results are already available in the literature. To overcome this problem, graphene oxide is functionalized by 1-aza-15-crown-5, the cage-like structure containing four oxygens that can bind Li(+) ion through electrostatic interaction. Li(+) migration on graphene surface has been investigated using ac relaxation mechanism. Perfect Debye-type relaxation behavior with β (relaxation exponent) value ≈1 resulting from single ion is observed. The activation energy of Li(+) migration arising due to cation-π interaction is found to be 0.37 eV, which agrees well with recently reported theoretical value. It is believed that this study will help to design isolated ion conductors for Li(+)-ion battery.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Lithium nitrate, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
1-Aza-15-crown-5, 97%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Lithium nitrate, BioUltra, ≥99.0% (calc. on dried substances, T)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol