Merck
  • Ultra-sensitive immunoassay biosensors using hybrid plasmonic-biosilica nanostructured materials.
Ultra-sensitive immunoassay biosensors using hybrid plasmonic-biosilica nanostructured materials.
Journal of biophotonics (2014-09-27)
Jing Yang, Le Zhen, Fanghui Ren, Jeremy Campbell, Gregory L Rorrer, Alan X Wang
ABSTRACT
We experimentally demonstrate an ultra-sensitive immunoassay biosensor using diatom biosilica with self-assembled plasmonic nanoparticles. As the nature-created photonic crystal structures, diatoms have been adopted to enhance surface plasmon resonances of metal nanoparticles on the surfaces of diatom frustules and to increase the sensitivity of surface-enhanced Raman scattering (SERS). In this study, a sandwich SERS immunoassay is developed based on the hybrid plasmonic-biosilica nanostructured materials that are functionalized with goat anti-mouse IgG. Our experimental results show that diatom frustules improve the detection limit of mouse IgG to 10 pg/mL, which is ˜100× better than conventional colloidal SERS sensors on flat glass. Ultra-sensitive immunoassay biosensor using diatom biosilica with self-assembled plasmonic nanoparticles.
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%