Merck
  • Simultaneous determination of acrylamide, asparagine and glucose in food using short chain methyl imidazolium ionic liquid based ultrasonic assisted extraction coupled with analyte focusing by ionic liquid micelle collapse capillary electrophoresis.
Simultaneous determination of acrylamide, asparagine and glucose in food using short chain methyl imidazolium ionic liquid based ultrasonic assisted extraction coupled with analyte focusing by ionic liquid micelle collapse capillary electrophoresis.
Food chemistry (2015-06-05)
Deia Abd El-Hady, Hassan M Albishri
ABSTRACT
Acrylamide (AA) is a known lethal neurotoxin and carcinogen. AA is formed in foods during the browning process by the Maillard reaction of glucose (GL) with asparagine (AS). For the first time, the simultaneous online preconcentration and separation of AA, AS and GL using analyte focusing by ionic liquid micelle collapse capillary electrophoresis (AFILMC) was presented. Samples were prepared in a 1-butyl-3-methylimidazolium bromide (BMIMBr) micellar matrix with a conductivity 4 times greater than that of the running buffer (12.5 mmol L(-1) phosphate buffer at pH 8.5). Samples were hydrodynamically injected into a fused silica capillary at 25.0 mbar for 25.0 s. Separations were performed by applying a voltage of 25.0 kV and a detection at 200.0 nm. To sufficiently reduce BMIMBr adsorption on the interior surface of capillary, an appropriate rinsing procedure by hydrochloric acid and water was optimized. AFILMC measurements of analytes within the concentration range of 0.05-10.0 μmol L(-1) achieved adequate reproducibility and accuracy with RSD 1.14-3.42% (n=15) and recovery 98.0-110.0%, respectively. Limits of detections were 0.71 ng g(-1) AA, 1.06 ng g(-1) AS and 27.02 ng g(-1) GL with linearity ranged between 2.2 and 1800 ng g(-1). The coupling of AFILMC with IL based ultrasonic assisted extraction (ILUAE) was successfully applied to the efficient extraction and determination of AA, AS and GL in bread samples. The structure of ILs has significant effects on the extraction efficiency of analytes. The optimal extraction efficiency (97.8%) was achieved by an aqueous extraction with 4:14 ratio of sample: 3.0 mol L(-1) BMIMBr followed by sonication at 35 °C. The proposed combination of ILUAE and AFILMC was simple, ecofriendly, reliable and inexpensive to analyze a toxic compound and its precursors in bread which is applicable to food safety.
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
L-Asparagine monohydrate, from non-animal source, BioReagent, suitable for cell culture, ≥98.0%