Merck
  • High efficiency polyethylene glycol diacrylate monoliths for reversed-phase capillary liquid chromatography of small molecules.
High efficiency polyethylene glycol diacrylate monoliths for reversed-phase capillary liquid chromatography of small molecules.
Journal of chromatography. A (2014-09-07)
Pankaj Aggarwal, John S Lawson, H Dennis Tolley, Milton L Lee
ABSTRACT
Highly cross-linked monolithic networks (i.e., polyethylene glycol diacrylate, PEGDA) synthesized from monomers containing varying ethylene oxide chain lengths were fabricated inside fused silica capillary columns for use in liquid chromatography (LC) of small molecules. Tergitol was used as a surfactant porogen in combination with other typical organic liquid porogens. Column performance was correlated with quantitative descriptors of the physical/chemical properties of the monomers and porogens using a statistical model. Solubility and viscosity values of the components were identified as important predictors of monolith morphology and efficiency. The chromatographic retention mechanism was determined to be principally reversed-phase (RP) with additional hydrogen bonding between the polar groups of the analytes and the ethylene oxide groups embedded in the monolith structure. The fabricated monolithic columns were evaluated under RPLC conditions using phenols, hydroxy benzoic acids, and alkyl parabens as test compounds. Isocratic elution of hydroxy benzoic acids at a linear velocity of 0.04 cm/s using a PEGDA-700 monolith gave chromatographic peaks with little tailing (i.e., tailing factor<1.28). The chromatographic efficiency measured for a non-retained compound (uracil) using this column was 186,000 plates/m when corrected for injector dead volume. High resolution gradient separations of selected pharmaceutical compounds and phenylurea herbicides were achieved in less than 18 min. Optimized monoliths synthesized from all four crosslinking monomers exhibited high permeability and demonstrated little swelling or shrinking in different polarity solvents. Column preparation was highly reproducible, with relative standard deviation (RSD) values less than 2.1%, based on retention times of the phenol standards (3 different columns).
Sigma-Aldrich
Resorcinol, ≥98%, FG
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Supelco
Linuron, PESTANAL®, analytical standard
Supelco
Diuron, PESTANAL®, analytical standard
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Resorcinol, meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
2,4-Dihydroxybenzoic acid, 97%
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%
Sigma-Aldrich
Resorcinol, BioXtra, ≥99%
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Diuron, ≥98%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Supelco
Isoproturon, PESTANAL®, analytical standard
Supelco
Ethylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Resorcinol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Propylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®