Merck
  • Injectable colloidal gold in a sucrose acetate isobutyrate gelating matrix with potential use in radiation therapy.
Injectable colloidal gold in a sucrose acetate isobutyrate gelating matrix with potential use in radiation therapy.
Advanced healthcare materials (2014-04-16)
Rasmus I Jølck, Tina Binderup, Anders E Hansen, Jonas B Scherman, Per Munch af Rosenschold, Andreas Kjaer, Thomas L Andresen
ABSTRACT
External beam radiation therapy relies on the ability to deliver high radiation doses to tumor cells with minimal exposure to surrounding healthy tissue. Advanced irradiation techniques, including image-guided radiation therapy (IGRT), rely on the ability to locate tumors to optimize the therapeutic benefit of these techniques. Today, radiopaque fiducial tissue markers are placed in or around tumors, for example, in prostate cancer patients to enhance the precision of daily and/or real-time IGRT. A liquid injectable fiducial marker (nanogel) is developed based on PEGylated gold nanoparticles and sucrose acetate isobutyrate (SAIB) with improved properties compared to current solid fiducial markers. The developed nanogel is investigated in vitro and subsequently evaluated in vivo in immunocompetent NMRI mice. The nanogel shows high CT-contrast and excellent stability in vivo over a period of 12 weeks. The nanogel is found to be biocompatible and well tolerated. No induction of the inflammatory cytokines INF-γ, IL-6, or TNF-α is observed throughout the study period. The developed nanogel seems to be a safe injectable fiducial marker ideally suited for IGRT that may further enhance the effect of radiation.
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Sucrose acetate isobutyrate
Sigma-Aldrich
Sodium citrate tribasic dihydrate, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Sodium citrate tribasic dihydrate, puriss. p.a., ACS reagent, ≥99.0% (NT)
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium citrate dihydrate, ≥99%, FG
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ≥98%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, for molecular biology, ≥99%
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Sodium citrate tribasic dihydrate, BioUltra, for molecular biology
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥65%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Sucrose acetate isobutyrate solution, 90 wt. % in denatured ethanol
Sigma-Aldrich
Trisodium citrate dihydrate, meets USP testing specifications