Merck
  • The guidance molecule Semaphorin3A is differentially involved in the arealization of the mouse and primate neocortex.
The guidance molecule Semaphorin3A is differentially involved in the arealization of the mouse and primate neocortex.
Cerebral cortex (New York, N.Y. : 1991) (2013-05-28)
Jihane Homman-Ludiye, James A Bourne
ABSTRACT
The visual cortex is organized into discrete domains characterized by their specific function, connectivity, chemoarchitecture, and cytoarchitecture. Gradients of transcription factors across the anteroposterior and mediolateral axes of the neocortex have previously been demonstrated to specify the main sensory regions. However, they do not account for the establishment of multiple areas in the primate visual cortex, which occupies approximately 50% of the neocortical surface. We demonstrate that the guidance molecule Semaphorin3A (Sema3A) is initially secreted in the cortical plate of the embryonic marmoset monkey and acts as an intrinsic cue to control the migration of subpopulations of neuronal progenitors and projection neurons expressing the receptor Neuropilin 1 (Npn1). During the first 2 postnatal weeks, Sema3A expression becomes primarily associated with ventral visual cortical areas, leading to the specific migration of Npn1+ neurons in the late maturing visual areas. In the mouse, Sema3A distribution is not arealized, but Npn1 expression becomes restricted to the posterior neocortex at embryonic day 16.5. The selective reduction in the striate cortex we observe in Sema3A-/- animals potentially results from the differential distribution of Npn1+ cells. Therefore, the Sema3A/Npn1 pathway participates to the parcellation of the visual neocortex in both the mouse and the marmoset, however, through different regulatory processes.
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Trometamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
SAFC
Tromethamine