Skip to Content
Merck
All Photos(4)

Key Documents

T6066

Sigma-Aldrich

Trizma® base

BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)

Synonym(s):

2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Beilstein:
741883
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.25

grade

BioPerformance Certified

Quality Level

Agency

USP/NF
meets EP testing specifications
meets USP testing specifications

description

aminopeptidase substrate

Assay

≥99.9% (titration)

form

crystalline powder

storage condition

dry at room temperature

technique(s)

DNA extraction: suitable
cell culture | mammalian: suitable
protein purification: suitable

impurities

DNase, RNase, protease, none detected
Endotoxin and Total Count Aerobic, Yeast, plus Mold, tested
≤0.2% water (Karl Fischer)

loss

≤0.5% (loss on drying, 110 °C)

color

white

pH

10.0-11.5

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)

mp

167-172 °C (lit.)

solubility

water: soluble (666 mg/ml, yielding a clear colorless solution)

absorption

≤0.05 at 260 at 1 M
≤0.05 at 290 at 40%
≤0.06 at 280 at 1 M

suitability

suitable for chromatography
suitable for immunoprecipitation
suitable for molecular biology

application(s)

clinical research
clinical testing
diagnostic assay manufacturing

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris base, also known as Trometamol or Trizma base, plays a crucial role in diverse research applications as a biological buffer. Widely utilized in buffer formulations like TAE and TBE buffers, its pKa of 8.1 renders it well-suited for maintaining pH within the physiological range (pH 7.0-9.0) applicable to most living organisms. Researchers, however, need to be cautious when studying proteins, as Tris may potentially interfere with the activity of certain enzymes.

This compound can be employed independently as a buffer or as part of mixed buffer formulations, including Tris-EDTA (TE) buffer, Tris-acetate-EDTA (TAE) buffer, Tris-borate-EDTA (TBE) buffer, among others. Notably, Tris base is pure, essentially stable, and relatively non-hygroscopic.

In laboratory settings, Tris base proves indispensable for preparing buffers compatible with biological fluids, serving as a standard pH solution, and facilitating various procedures such as lactate dehydrogenase assays, in situ hybridization, and protein extraction from cells. Its versatility extends to applications in cell culture, biochemistry, and molecular biology labs, where it aids in studies related to cell membrane permeability and buffer preparation.

Application

Trizma has been used:
  • As a component in the culture of human embryonic stem (hES) cells on human and mouse feeder cells
  • As a component of pre-hybridization buffer in in situ hybridization
  • To suspend dried protein before SDS-PAGE protein characterization
  • As a component of lysis buffer to wash and lyse cells before immunoprecipitation
  • In the extraction and enrichment of membrane proteins for separation by two-dimensional polyacrylamide gel electrophoresis
  • In studies of double stranded complexes of peptide nucleic acids (PNA) and their complementary DNA sequences, by use of anion exchange HPLC
  • In capillary electrochromatography and UV analysis of tocopherols and tocotrienols
  • as a component of Tris/base solution in a sulforhodamine B (SRB) assay

Features and Benefits

  • Suitable for Cell culture, Biochemical and Molecular Biology Research
  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications
  • Effective Buffering from pH 7-9 (25 °C) with a pKa of 8.1 (25 °C)
  • Tested for the presence of Anionic and Cationic traces
  • Tested for DNase, RNase, and protease

Packaging

T6066-1KT:
Each kit contains 3 x 100G samples, each sample from a uniquely manufactured lot.

Other Notes

The pH values of all buffers are temperature- and concentration-dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution.
For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.
For additional information on our range of Biochemicals, please complete this form.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Culture of human embryonic stem cells on human and mouse feeder cells.
Turksen, Kursad
Human Embryonic Stem Cell Protocols, 91-104 (2006)
Recurrent mutations in the MTOR regulator RRAGC in follicular lymphoma.
Peterson LF, et al.
Clinical Cancer Research, 22(21), 5383-5393 (2016)
Membrane proteins and proteomics: un amour impossible?
Santoni V, et al.
Electrophoresis, 21(6), 1054-1070 (2000)
Capillary electrochromatographic evaluation of vitamin active oil constituents: tocopherols and tocotrienols.
Abidi SL and Rennick KA
Journal of Chromatography A, 913(1-2), 379-386 (2001)
Recognition and strand displacement of DNA oligonucleotides by peptide nucleic acids (PNAs). High-performance ionexchange chromatographic analysis
Lesignoli F, et al.
Journal of Chromatography A, 922(1-2), 177-185 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service