Merck
  • Meta-analysis to assess the efficacy and toxicity of docetaxel-based doublet compared with docetaxel alone for patients with advanced NSCLC who failed first-line treatment.
Meta-analysis to assess the efficacy and toxicity of docetaxel-based doublet compared with docetaxel alone for patients with advanced NSCLC who failed first-line treatment.
Clinical therapeutics (2014-09-27)
Ying Jin, Yan Sun, Xun Shi, Jun Zhao, Lei Shi, Wei Hong, Xinmin Yu
ABSTRACT
The benefit of docetaxel-based therapy in the second-line treatment of advanced non-small cell lung cancer (NSCLC) is still unclear. The goal of this meta-analysis was to assess the efficacy and toxicity of docetaxel-based doublet compared with docetaxel alone for patients with advanced NSCLC who failed to improve with first-line treatment. Several databases were searched, including PubMed, Embase, and the Cochrane databases. The end points were overall survival, progression-free survival (PFS), objective response rate, disease control rate, and grade 3 or 4 adverse events. Data were extracted from the studies by 2 independent reviewers. The meta-analysis was performed by using Review Manager version 5.2. The pooled hazard ratio (HR) or odds ratio (OR) and 95% CIs were calculated by using fixed or random effects models depending on the heterogeneity of the included trials. Twelve eligible trials involving 2680 patients were identified. The intention-to-treatment analysis found that docetaxel-based therapy significantly improved overall survival (HR, 0.89 [95% CI, 0.83-0.96]; P < 0.01), PFS (HR, 0.79 [95% CI, 0.71-0.89]; P < 0.01), objective response rate (OR, 1.73 [95% CI, 1.37-2.18; P < 0.01), and disease control rate (OR, 1.30 [95% CI, 1.09-1.55]; P < 0.01). In addition, a subgroup analysis based on type of combined drug showed that there were significant improvement in PFS and overall survival in combining docetaxel with targeted therapy. In addition, a higher incidence of grade 3 or 4 diarrhea and thrombocytopenia was observed in docetaxel-based doublet therapy. Based on the available evidence, docetaxel-based doublet therapy seems superior to docetaxel monotherapy as a second-line treatment for advanced NSCLC. More studies should focus on combining docetaxel with targeted therapy to identify patients who will most likely benefit from the appropriate combination targeted therapy.
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Anhydrous Docetaxel, European Pharmacopoeia (EP) Reference Standard
USP
Docetaxel, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ifosfamide, ≥98%
Sigma-Aldrich
Hydriodic acid, 57 wt. %, distilled, 99.999% trace metals basis
Ifosfamide, European Pharmacopoeia (EP) Reference Standard
USP
Ifosfamide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydriodic acid, contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, distilled, 57 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Hydriodic acid, 57 wt. % in H2O, distilled, stabilized, 99.95%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, ACS reagent, 55%