Merck
  • Selectivity of gas-phase ion/molecule reaction of carbon dioxide with phenide ions.
Selectivity of gas-phase ion/molecule reaction of carbon dioxide with phenide ions.
Journal of mass spectrometry : JMS (2014-07-22)
Chongming Liu, Yong Zhang, Athula B Attygalle
ABSTRACT
On contrary to the widely accepted conviction that the m/z 93 ion derived from phenol does not react with CO2, we demonstrate that it makes an adduct with CO2 to a small but demonstrable extent. For example, the product-ion mass spectrum recorded for the m/z 98 ion derived from [(2)H6]phenol showed a small peak at m/z 142 when CO2 was used as the collision gas. The formation of an m/z 137 adduct ion from the m/z 93 ion (generated either directly from phenol, or indirectly from salicylic acid by in-source decarboxylation) was demonstrated also by multiple-reaction-monitoring tandem mass spectrometric experiments. According to literature, the m/z 93 ion derived from salicylic acid does not undergo CO2 addition because it is deemed to exist only in the phenoxide form. This reaction has been previously proposed as a method for differentiating phenoxide ion from its isomeric hydroxyphenide ions. We propose that the m/z 93 ion, albeit small, exists also as the phenide form together with the predominant phenoxide ion.
Product Description
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Mesalazine impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phenol, natural, 97%, FG
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
4-Methoxyphenol, purum, ≥98.0% (HPLC)
Supelco
4-Aminophenol, PESTANAL®, analytical standard
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
4-Aminophenol, ≥98%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)