Merck
  • Corneal cross-linking in keratoconus using the standard and rapid treatment protocol: differences in demarcation line and 12-month outcomes.
Corneal cross-linking in keratoconus using the standard and rapid treatment protocol: differences in demarcation line and 12-month outcomes.
Investigative ophthalmology & visual science (2014-12-04)
Sara Brittingham, Christoph Tappeiner, Beatrice E Frueh
ABSTRACT
To compare the occurrence rate and depth of the demarcation line and topographical outcome after corneal cross-linking (CXL) for keratoconus using two different treatment protocols. A retrospective analysis of 131 eyes with progressive keratoconus treated with CXL using riboflavin and UV-A was performed. Eyes were treated either with the standard Dresden protocol (30 minutes irradiation, 3 mW/cm(2), UV-XTM 1000) or a rapid protocol (10 minutes irradiation, 9 mW/cm(2), UV-XTM 2000). The presence and depth of the corneal demarcation line was assessed with an anterior segment optical coherence tomography device 1 month after CXL by a masked observer. Corneal topography and tomography was performed at baseline and at 12-month follow-up with Pentacam and the TMS (Topographic Modeling System) device. In the standard protocol group, 76.5% (62/81) of treated corneas revealed a demarcation line 1 month after CXL, whereas such a demarcation line was observed in only 22% (11/50) of eyes treated with the rapid protocol (P < 0.0001). The demarcation line was significantly more superficial in the rapid protocol group (P = 0.004). Corneal topography values between baseline and 12 months after CXL showed a mean change of -0.76 diopters (D) in Kmax (SD ± 2.7) in the standard protocol group versus a mean change of +0.72 D in Kmax (SD ± 1.5) in the rapid protocol (P = 0.007). The rapid CXL protocol negatively influences the occurrence and depth of the demarcation line 1 month after CXL. Our results show a negative effect on the topographical outcome 1 year after CXL.
Product Description
Supelco
Tetracaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetracaine hydrochloride, ≥99%
Sigma-Aldrich
Fluorometholone, ≥98%
USP
Fluorometholone, United States Pharmacopeia (USP) Reference Standard
Tetracaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Tetracaine for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Tetracaine hydrochloride, United States Pharmacopeia (USP) Reference Standard