Skip to Content
Merck
All Photos(1)

Key Documents

1278007

USP

Fluorometholone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

11β,17α-Dihydroxy-9-fluoro-6-methyl-1,4-pregnadiene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29FO4
CAS Number:
Molecular Weight:
376.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fluorometholone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(C)=O)[C@@]3(C)C[C@H](O)[C@]2(F)[C@@]4(C)C=CC(=O)C=C14

InChI

1S/C22H29FO4/c1-12-9-17-15-6-8-21(27,13(2)24)20(15,4)11-18(26)22(17,23)19(3)7-5-14(25)10-16(12)19/h5,7,10,12,15,17-18,26-27H,6,8-9,11H2,1-4H3/t12-,15-,17-,18-,19-,20-,21-,22-/m0/s1

InChI key

FAOZLTXFLGPHNG-KNAQIMQKSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluorometholone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Fluorometholone Acetate
  • Fluorometholone Ophthalmic Suspension
  • Hydrocortisone Hemisuccinate
  • Hydrocortisone Sodium Succinate for Injection
  • Methylprednisolone Hemisuccinate
  • Methylprednisolone Sodium Succinate for Injection

Biochem/physiol Actions

Clinically significant in allergic conjunctivitis and as anti-inflammatory following cataract surgery.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua Gao et al.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie, 252(6), 963-968 (2014-05-07)
To evaluate the feasibility of partial lamellar keratoplasty (LK) for treatment of peripheral corneal disease (PCD) using a graft from the corneoscleral rim preserved in glycerin. Patients who underwent LK for PCD at Shandong Eye Hospital from January 2006 to
Asimina Mataftsi et al.
The British journal of ophthalmology, 95(11), 1531-1533 (2011-02-08)
Although fluorometholone (FML) is considered a steroid of minimal ocular penetration, reports in children have shown dose-dependent intraocular pressure (IOP) rise. The authors aimed to assess whether reducing regimens of FML for paediatric ocular surface disease have sustained clinically significant
Wenjia Xie et al.
Investigative ophthalmology & visual science, 55(9), 5806-5812 (2014-08-26)
To investigate the effect on tear menisci after laser in situ keratomileusis (LASIK) with flap creation by either microkeratome or femtosecond laser. Sixty eyes of 30 myopes were analyzed. Fifteen patients underwent LASIK with Moria II microkeratome, and the other
Miaomiao Zhang et al.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie, 252(6), 983-987 (2014-03-29)
To analyze the efficacy of selective laser trabeculoplasty (SLT) on silicone oil-induced secondary glaucoma in terms of intraocular pressure (IOP). 42 patients (42 eyes) with silicone oil-induced secondary glaucoma were selected, and SLT was performed with 360° of the trabecular
Dong Hui Lim et al.
American journal of ophthalmology, 158(3), 447-454 (2014-06-22)
To investigate visual outcomes of Visian implantable Collamer lens (ICL) implantation in eyes with a shallow anterior chamber depth (ACD). Retrospective, noncomparative, interventional case series. Medical charts of the patients with a low ACD of 2.8 mm or less who

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service