Merck
  • Methylene blue modulates adhesion molecule expression on microvascular endothelial cells.
Methylene blue modulates adhesion molecule expression on microvascular endothelial cells.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2014-05-06)
Isabella Werner, Fengwei Guo, Ulrich A Stock, Michèle Lupinski, Patrick Meybohm, Anton Moritz, Andres Beiras-Fernandez
ABSTRACT
As methylene blue (MB) has been recently proposed to preserve blood pressure in case of vasoplegic syndrome and shock, an entity directly related to systemic inflammation, we aimed to elucidate the effect of MB on the expression of adhesion-molecules in endothelial-cells. Human microvascular endothelial-cells (HuMEC-1) were treated with 10, 30 or 60 µM MB for 30 min and 2 h each. Additionally, the treated HuMEC-1 were co-cultured with either human peripheral blood mononuclear cells (PBMCs) or Jurkat cells (human T-lymphocytes) for 2 h. HuMEC-1 were analyzed after MB treatment and after co-culture experiments for expression of different adhesion-molecules (ICAM-1, VCAM-1, L-selectin, E-selectin) via FACS measurement and western blot analysis. The supernatants of the experiments were analyzed with regard to the soluble forms of the adhesion molecules. We found that MB is able to modulate the expression of adhesion-molecules on EC. Administration of MB increases the expression of E-selectin and VCAM-1 depending on the dosage and time of exposure. ICAM-1 measurements provide evidence that different circulating blood cells can differently alter the adhesion-molecule expression on EC after MB exposure. Our results provide evidence regarding the immunomodulatory effect of MB upon endothelial-cells after inflammation.
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Human VCAM-1 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Mouse ICAM-1 ELISA Kit, for serum, plasma and cell culture supernatant
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Human E-Selectin ELISA Kit, for serum, plasma, cell culture supernatants and urine
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Human sICAM1 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Rat ICAM-1 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Mouse VCAM-1 ELISA Kit, for serum, plasma, cell culture supernatant
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)