Merck
  • Biological activities and phytochemical profiles of extracts from different parts of bamboo (Phyllostachys pubescens).
Biological activities and phytochemical profiles of extracts from different parts of bamboo (Phyllostachys pubescens).
Molecules (Basel, Switzerland) (2014-06-20)
Akinobu Tanaka, Qinchang Zhu, Hui Tan, Hiroki Horiba, Koichiro Ohnuki, Yasuhiro Mori, Ryoko Yamauchi, Hiroya Ishikawa, Akira Iwamoto, Hiroharu Kawahara, Kuniyoshi Shimizu
ABSTRACT
Besides being a useful building material, bamboo also is a potential source of bioactive substances. Although some studies have been performed to examine its use in terms of the biological activity, only certain parts of bamboo, especially the leaves or shoots, have been studied. Comprehensive and comparative studies among different parts of bamboo would contribute to a better understanding and application of this knowledge. In this study, the biological activities of ethanol and water extracts from the leaves, branches, outer culm, inner culm, knots, rhizomes and roots of Phyllostachys pubescens, the major species of bamboo in Japan, were comparatively evaluated. The phytochemical profiles of these extracts were tentatively determined by liquid chromatography-mass spectrometry (LC-MS) analysis. The results showed that extracts from different parts of bamboo had different chemical compositions and different antioxidative, antibacterial and antiallergic activities, as well as on on melanin biosynthesis. Outer culm and inner culm were found to be the most important sources of active compounds. 8-C-Glucosylapigenin, luteolin derivatives and chlorogenic acid were the most probable compounds responsible for the anti-allergy activity of these bamboo extracts. Our study suggests the potential use of bamboo as a functional ingredient in cosmetics or other health-related products.
Product Description
Chlorogenic acid, primary reference standard
Sigma-Aldrich
Interleukin-4 human, IL-4, potency:, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
IL-4 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
SOD Assay Kit, sufficient for 500 tests
Sigma-Aldrich
IL-4 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Syringic acid, ≥95% (HPLC)
Sigma-Aldrich
Interleukin-4 human, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
trans-Ferulic acid, 99%
Ferulic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Syringic acid, analytical standard