Merck
  • Mutagenicity and DNA-damaging potential of clenbuterol and its metabolite 4-amino-3,5-dichlorobenzoic acid in vitro.
Mutagenicity and DNA-damaging potential of clenbuterol and its metabolite 4-amino-3,5-dichlorobenzoic acid in vitro.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2015-01-18)
Ana Vulić, Ksenija Durgo, Jelka Pleadin, Luka Herceg, Nevenka Kopjar
ABSTRACT
The aim of this study was to evaluate in vitro toxicity of clenbuterol and its metabolite 4-amino-3,5-dichlorobenzoic acid. Cytotoxicity and pro-oxidative effect of both compounds were studied on human colon adenocarcinoma cell line SW 480. No significant cytotoxic effect of either compound was observed. Results of an Ames test on Salmonella typhimurium did not indicate mutagenic activity of clenbuterol on TA 98 and TA 100 strains, regardless of metabolic activation. Potential mutagenic effects of the highest clenbuterol concentration (2500 ng/ml) were observed on the TA 1535 strain. The obtained results of alkaline comet assay on isolated human lymphocytes suggested that both compounds induced an increase of primary DNA damage in a concentration-dependent manner. 4-ADBA was a slightly more potent inducer of primary DNA damage as compared to clenbuterol. Chromosomal aberration analysis showed that clenbuterol caused a statistically significant increase in the total number of aberrant cells only at the highest concentration tested (3% vs. 0.7% in the negative control). The results of this study might represent a solid frame for designing and planning future studies with both compounds, which should further clarify their mechanisms of action and genotoxic/cytogenetic effects relevant for human risk assessment.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, ≥97.0% (HPLC)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, BioUltra, for molecular biology, ≥99.0% (HPLC)
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, ≥94%
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, detergent for use in cell lysis
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, BioXtra, ≥97% (TLC)
SAFC
HEPES
Supelco
9-Aminoacridine, matrix substance for MALDI-MS, ≥99.5% (HPLC)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%