Merck
  • Vasopressin-regulated miRNAs and AQP2-targeting miRNAs in kidney collecting duct cells.
Vasopressin-regulated miRNAs and AQP2-targeting miRNAs in kidney collecting duct cells.
American journal of physiology. Renal physiology (2015-02-05)
Jae-Eun Kim, Hyun Jun Jung, Yu-Jung Lee, Tae-Hwan Kwon
ABSTRACT
Mature microRNA (miRNA) acts as an important posttranscriptional regulator. We aimed to profile vasopressin-responsive miRNAs in kidney inner medullary collecting duct cells and to identify aquaporin-2 (AQP2)-targeting miRNAs. Microarray chip assay was carried out in inner medullary collecting duct tubule suspensions from rat kidneys in the absence or presence of desmopressin (dDAVP) stimulation (10(-9) M, 2 h). The results demonstrated 19 miRNAs, including both precursor and mature miRNAs, as potential candidates that showed significant changes in expression after dDAVP stimulation (P < 0.05). Nine mature miRNAs exhibiting >1.3-fold changes in expression on the microarray (miR-127, miR-1, miR-873, miR-16, miR-206, miR-678, miR-496, miR-298, and miR-463) were further examined by quantitative real-time PCR, and target genes of the selected miRNAs were predicted. Next, to identify AQP2-targeting miRNAs, in silico analysis was performed. Four miRNAs (miR-32, miR-137, miR-216a, and miR-216b) target the 3'-untranslated region of rat AQP2 mRNA. Target seed regions of miR-32 and miR-137 were also conserved in the 3'-untranslated region of mouse AQP2 mRNA. Quantitative real-time PCR and immunoblot analysis demonstrated that dDAVP-induced AQP2 expression was significantly attenuated in mpkCCDc14 cells when cells were transfected with miRNA mimics of miR-32 or miR-137. Moreover, luciferase reporter assay demonstrated a significant decrease of AQP2 translation in mpkCCDc14 cells transfected with miRNA mimics of miR-32 or miR-137. The present study provides novel insights into the regulation of AQP2 by RNA interference; however, vasopressin-regulated miRNAs did not include miR-32 or miR-137, indicating that the interaction of miRNAs with the AQP2 regulatory pathway requires further analysis.
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
Guanine, BioUltra
Sigma-Aldrich
S-(5′-Adenosyl)-L-homocysteine, crystalline
Supelco
Guanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Anti-Aquaporin 2 Antibody, Chemicon®, from rabbit