Merck
  • The C3G/Rap1 pathway promotes secretion of MMP-2 and MMP-9 and is involved in serous ovarian cancer metastasis.
The C3G/Rap1 pathway promotes secretion of MMP-2 and MMP-9 and is involved in serous ovarian cancer metastasis.
Cancer letters (2015-01-27)
Ya-Ling Che, Shu-Juan Luo, Gang Li, Min Cheng, Yi-Meng Gao, Xue-Mei Li, Jie-Min Dai, Huan He, Jin Wang, Hui-Juan Peng, Yu Zhang, Wen-Yan Li, Hui Wang, Bin Liu, Hua Linghu
ABSTRACT
Complete resection is pivotal to improve survival to epithelial ovarian cancer (EOC). Crk SH3-domain-binding guanine nucleotide-releasing factor (C3G) is involved in multiple signaling pathways and it has opposite roles in different cancers. The present study aimed to identify C3G expression in ovarian tissue samples from patients with EOC and to explore its association with tumor grade. Eighty-seven archival paraffin-embedded, formalin-fixed, ovarian cancer tissues with serous histology were stained for C3G by immunohistochemistry. To evaluate the contribution of C3G to Rap1 activity, 36 patients with serous ovarian cancer (SOC) were investigated. Additionally, C3G was knocked down in SKOV3 and HEY cells. C3G regulated Rap1 activity and high Rap1 activity was correlated with poor differentiation, advanced FIGO stage, and unsuccessful cytoreductive surgery of SOC. Knockdown of C3G suppressed cell invasion, intravasation and extravasation, and reduced Rap1 activity and secretion of matrix metalloproteinase (MMP)-2 and MMP-9. C3G-mediated activation of Rap1 could direct the tumor pattern of human SOC by promoting the secretion of MMP-2 and MMP-9. These results suggest that C3G is involved in the metastatic spread of EOC.
Product Description
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, for ultratrace analysis, 30 % (w/w)
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Supelco
o-Xylene, analytical standard
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Supelco
o-Xylene, suitable for HPLC, 98%
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%